Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HBB-BS SIRNA Assays ›
          Gene Symbol:
          Hbb-bs Hbb-b1 Hbb-b2 Hbb-bt
          Gene Name:
          hemoglobin, beta adult s chain
          hemoglobin, beta adult major chain
          hemoglobin, beta adult minor chain
          hemoglobin, beta adult t chain
          Gene Aliases:
          Beta-s, Hbbt1, Hbbt2
          AA409645, Hbb1, Hbbt1, Hbbt2, MommeD7, beta1
          AI036344, Hbb2, Hbbt2, beta2
          Beta-t
          Chromosome Location:
          Chr.7: 103826523 - 103827928 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          -
          Chromosome Location:
          Chr.7: 103812524 - 103813923 on Build GRCm38
          UniGene ID:
          Mm.288567
          Mm.467412
          Mm.288567
          Mm.467412
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s231963

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hbb-bs
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001201391.1 NP_001188320.1 2 179
          GenBank mRNA AK002258.1 - 2 179
          AK003096.1 - 2 179
          AK003472.1 - 2 179
          AK005442.1 - 2 179
          AK005490.1 - 2 180
          AK010873.1 - 2 180
          AK010902.1 - 2 179
          AK010980.1 - 2 179
          AK010981.1 - 2 179
          AK010991.1 - 2 179
          AK010993.1 - 2 178
          AK011006.1 - 2 179
          AK011013.1 - 2 179
          AK011016.1 - 2 179
          AK011027.1 - 2 179
          AK011053.1 - 2 181
          AK011067.1 - 2 179
          AK011069.1 - 2 181
          AK011077.1 - 2 179
          AK011083.1 - 2 179
          AK011102.1 - 2 179
          AK027903.1 - 2 181
          AK027904.1 - 2 179
          AK088149.1 - 2 179
          AK133714.1 - 2 179
          AK147001.1 - 2 181
          AK160629.1 - 2 179
          AK161021.1 - 2 179
          AK165490.1 - 2 179
          AK168562.1 - 2 180
          AK168826.1 - 2 180
          AK168846.1 - 2 179
          Gene Symbol: Hbb-b1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001278161.1 NP_001265090.1 2 180
          GenBank mRNA AB020015.1 - 2 123
          M10524.1 - 2 176
          S71555.1 - 2 239
          S71558.1 - 2 239
          S71559.1 - 2 239
          Gene Symbol: Hbb-b2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_016956.3 NP_058652.1 2 180
          GenBank mRNA AB020014.1 - 2 123
          AB020016.1 - 2 123
          AB020017.1 - 2 123
          AB364478.1 - 2 126
          AF149782.1 - 2 126
          BC027434.1 - 2 160 4920093
          BC032264.1 - 2 171 3988455
          Gene Symbol: Hbb-bt
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008220.5 NP_032246.2 2 180
          GenBank mRNA AK002394.1 - 2 179
          AK005496.1 - 2 179
          AK011033.1 - 2 179
          AK011049.1 - 194
          AK011050.1 - 2 180
          AK011052.1 - 2 179
          AK011057.1 - 2 179
          AK011075.1 - 89
          AK012551.1 - 2 179
          AK014364.1 - 2 180
          AK028067.1 - 2 182
          AK167615.1 - 2 179
          AK168412.1 - 2 178
          AK168477.1 - 2 179
          AK168584.1 - 2 179
          AK168819.1 - 2 179

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of eIF2 alpha phosphorylation by heme
          oxygen transport
          hemopoiesis
          erythrocyte development
          transport
          positive regulation of myeloid cell differentiation
          regulation of erythrocyte differentiation
          peroxidase activity
          oxygen binding
          hemoglobin binding
          haptoglobin binding
          hemoglobin alpha binding
          hemoglobin beta binding
          oxygen transporter activity
          iron ion binding
          heme binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-wtw7d:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0