Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRP6 SIRNA Assays ›
Gene Symbol:
Lrp6
Gene Name:
low density lipoprotein receptor-related protein 6
Gene Aliases:
C030016K15Rik, Cd, Gw, ska26, ska<m26Jus>, skax26
Chromosome Location:
Chr.6: 134446476 - 134566960 on Build GRCm38
UniGene ID:
Mm.321990
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
s232058
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
404.00
Your Price
Online offer:
391.65
391.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Lrp6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008514.4
NP_032540.2
11, 12
2688
GenBank mRNA
AF074265.1
-
11, 12
2557
BC060704.1
-
11, 12
2688
6409058
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107892, XM_006237519.
Panther Classification:
Molecular Function -
apolipoprotein
Gene Ontology Categories:
Function(s)
Process(es)
gastrulation with mouth forming second
somitogenesis
neural tube closure
negative regulation of protein phosphorylation
heart looping
morphogenesis of an epithelium
positive regulation of mesenchymal cell proliferation
pericardium morphogenesis
axis elongation
regulation of transcription, DNA-templated
negative regulation of protein kinase activity
endocytosis
receptor-mediated endocytosis
positive regulation of cytosolic calcium ion concentration
chemical synaptic transmission
multicellular organism development
embryonic pattern specification
dorsal/ventral axis specification
anterior/posterior pattern specification
neural crest formation
neural crest cell differentiation
Wnt signaling pathway
single organismal cell-cell adhesion
cerebellum morphogenesis
thalamus development
cerebral cortex cell migration
forebrain radial glial cell differentiation
forebrain generation of neurons
Wnt signaling pathway involved in forebrain neuroblast division
neural tube development
formation of radial glial scaffolds
cerebral cortex development
negative regulation of Wnt signaling pathway
embryonic limb morphogenesis
forebrain development
midbrain development
midbrain-hindbrain boundary development
negative regulation of smooth muscle cell apoptotic process
limb morphogenesis
embryonic forelimb morphogenesis
embryonic hindlimb morphogenesis
external genitalia morphogenesis
post-anal tail morphogenesis
cell migration involved in gastrulation
regulation of cell proliferation
odontogenesis of dentin-containing tooth
embryonic digit morphogenesis
positive regulation of apoptotic process
Wnt signaling pathway involved in dorsal/ventral axis specification
negative regulation of fat cell differentiation
positive regulation of ossification
positive regulation of bone resorption
positive regulation of cell cycle
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
bone remodeling
embryonic camera-type eye morphogenesis
generation of neurons
skeletal system morphogenesis
negative regulation of epithelial cell proliferation
positive regulation of sequence-specific DNA binding transcription factor activity
response to folic acid
palate development
convergent extension
retina morphogenesis in camera-type eye
embryonic retina morphogenesis in camera-type eye
canonical Wnt signaling pathway
regulation of cell development
face morphogenesis
bone morphogenesis
branching involved in mammary gland duct morphogenesis
trachea cartilage morphogenesis
mammary placode formation
mammary gland duct morphogenesis
canonical Wnt signaling pathway involved in cardiac neural crest cell differentiation involved in heart development
canonical Wnt signaling pathway involved in positive regulation of cardiac outflow tract cell proliferation
cellular response to cholesterol
dopaminergic neuron differentiation
negative regulation of protein serine/threonine kinase activity
primitive streak formation
receptor-mediated endocytosis of low-density lipoprotein particle involved in cholesterol transport
Wnt signaling pathway involved in somitogenesis
axis elongation involved in somitogenesis
positive regulation of canonical Wnt signaling pathway
toxin transport
negative regulation of non-canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway involved in dorsal/ventral axis specification
negative regulation of planar cell polarity pathway involved in ventricular septum morphogenesis
negative regulation of planar cell polarity pathway involved in cardiac muscle tissue morphogenesis
negative regulation of planar cell polarity pathway involved in cardiac right atrium morphogenesis
negative regulation of planar cell polarity pathway involved in outflow tract morphogenesis
negative regulation of planar cell polarity pathway involved in pericardium morphogenesis
negative regulation of planar cell polarity pathway involved in neural tube closure
low-density lipoprotein receptor activity
receptor binding
frizzled binding
protein binding
Wnt-protein binding
kinase inhibitor activity
toxin transporter activity
apolipoprotein binding
identical protein binding
protein homodimerization activity
Wnt-activated receptor activity
coreceptor activity involved in Wnt signaling pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0