Search
Search

Nanog| Gene Symbol: Nanog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289828.1 | NP_001276757.1 | 4 | 1159 | ||
| NM_001289830.1 | NP_001276759.1 | 5 | 959 | |||
| NM_001289831.1 | NP_001276760.1 | 5 | 943 | |||
| NM_028016.3 | NP_082292.1 | 4 | 1162 | |||
| GenBank mRNA | AB093574.1 | - | 4 | 1133 | ||
| AF507043.1 | - | 4 | 1134 | |||
| AK010332.1 | - | 4 | 1134 | |||
| AK167259.1 | - | 4 | 1134 | |||
| AY278951.1 | - | 4 | 1011 | |||
| AY455282.1 | - | 5 | 943 | |||
| AY455285.1 | - | 5 | 959 | |||
| BC083105.1 | - | 1 | 42 | |||
| BC132017.1 | - | 4 | 1159 | 40130294 | ||
| BC137873.1 | - | 4 | 1159 | 8860892 | ||
| BC144995.1 | - | 4 | 1156 | 9053543 | ||
| BC144996.1 | - | 4 | 1159 | 9053544 | ||