Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KANK1 SIRNA Assays ›
          Gene Symbol:
          KANK1
          Gene Name:
          KN motif and ankyrin repeat domains 1
          Gene Aliases:
          ANKRD15, CPSQ2, KANK
          Chromosome Location:
          Chr.9: 470294 - 746106 on Build GRCh38
          UniGene ID:
          Hs.306764
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23236

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KANK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001256876.1 NP_001243805.1 8, 9 3553
          NM_001256877.1 NP_001243806.1 5, 6 3662
          NM_015158.3 NP_055973.2 4, 5 3047
          NM_153186.4 NP_694856.1 3, 4 3309
          XM_005251411.2 XP_005251468.1 4, 5 3047
          XM_005251414.3 XP_005251471.1 3, 4 3510
          XM_005251415.2 XP_005251472.1 3, 4 3239
          XM_005251416.2 XP_005251473.1 3, 4 3015
          XM_005251417.3 XP_005251474.1 3, 4 3506
          XM_005251418.2 XP_005251475.1 3, 4 3517
          XM_005251419.2 XP_005251476.1 3, 4 3208
          XM_006716743.2 XP_006716806.1 4, 5 3023
          XM_011517819.1 XP_011516121.1 5, 6 3138
          XM_011517820.1 XP_011516122.1 4, 5 3155
          XM_011517821.1 XP_011516123.1 4, 5 3082
          XM_017014511.1 XP_016870000.1 5, 6 3260
          XM_017014512.1 XP_016870001.1 4, 5 4571
          XM_017014513.1 XP_016870002.1 6, 7 3773
          XM_017014514.1 XP_016870003.1 4, 5 3459
          XM_017014515.1 XP_016870004.1 5, 6 3195
          XM_017014519.1 XP_016870008.1 4, 5 3459
          XM_017014520.1 XP_016870009.1 4, 5 3047
          XM_017014521.1 XP_016870010.1 4, 5 3459
          XM_017014522.1 XP_016870011.1 5, 6 3260
          XM_017014523.1 XP_016870012.1 4, 5 3047
          XM_017014526.1 XP_016870015.1 4, 5 3492
          XM_017014527.1 XP_016870016.1 3, 4 3239
          XM_017014528.1 XP_016870017.1 4, 5 3093
          XM_017014529.1 XP_016870018.1 3, 4 3506
          XM_017014530.1 XP_016870019.1 3, 4 3015
          XM_017014533.1 XP_016870022.1 3, 4 2956
          XM_017014537.1 XP_016870026.1 3, 4 3028
          XM_017014538.1 XP_016870027.1 3, 4 3513
          XM_017014539.1 XP_016870028.1 3, 4 3028
          XM_017014540.1 XP_016870029.1 3, 4 3504
          XM_017014541.1 XP_016870030.1 3, 4 3508
          GenBank mRNA AK025428.1 - 2, 3 1192
          AK026421.1 - 2, 3 2662
          AK290028.1 - 4, 5 3013
          AK292989.1 - 4, 5 3026
          AL833161.1 - 4, 5 3027
          BC020040.1 - 2, 3 1201
          BC037495.2 - 8, 9 3556 5265517
          BC038116.2 - 3, 4 3219 5261060
          BX648931.1 - 4, 5 3041
          D79994.2 - 4, 5 3612

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of neuron projection development
          positive regulation of Wnt signaling pathway
          negative regulation of cell migration
          negative regulation of actin filament polymerization
          negative regulation of Rho protein signal transduction
          positive regulation of catenin import into nucleus
          negative regulation of insulin receptor signaling pathway
          positive regulation of wound healing
          negative regulation of substrate adhesion-dependent cell spreading
          negative regulation of ruffle assembly
          regulation of establishment of cell polarity
          negative regulation of lamellipodium morphogenesis
          protein binding
          beta-catenin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5rpk4:80/100.66.77.5:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0