Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Rab15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134050.4 | NP_598811.3 | 2 | 345 | ||
GenBank mRNA | AB232610.1 | - | 2 | 138 | ||
AK158637.1 | - | 2 | 267 | |||
AK158768.1 | - | 2 | 299 | |||
BC013790.1 | - | 2 | 291 | 2655151 | ||
BC027769.1 | - | 2 | 345 | 5345297 | ||
BY237353.1 | - | 2 | 346 |