Search
Search

Asap1| Gene Symbol: Asap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276461.1 | NP_001263390.1 | 7, 8 | 752 | ||
| NM_001276462.1 | NP_001263391.1 | 5, 6 | 661 | |||
| NM_001276463.1 | NP_001263392.1 | 5, 6 | 661 | |||
| NM_001276467.1 | NP_001263396.1 | 7, 8 | 752 | |||
| NM_010026.3 | NP_034156.2 | 7, 8 | 752 | |||
| GenBank mRNA | AF075461.1 | - | 7, 8 | 571 | ||
| AF075462.1 | - | 7, 8 | 571 | |||
| AK122477.1 | - | 5, 6 | 356 | |||
| AK132979.1 | - | 491 | ||||
| AK146545.1 | - | 7, 8 | 590 | |||
| AK147689.1 | - | 5, 6 | 662 | |||
| BC048818.1 | - | 5, 6 | 627 | |||
| BC094581.1 | - | 7, 8 | 752 | 30549241 | ||
| CB527210.1 | - | 5, 6 | 455 | |||
| CD355387.1 | - | 3, 4 | 249 | |||