Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103179.1 | NP_001096649.1 | 5 | 391 | ||
NM_145125.3 | NP_660107.2 | 5 | 391 | |||
NM_176928.1 | NP_795902.1 | 5 | 391 | |||
GenBank mRNA | AJ292467.1 | - | 5 | 396 | ||
AJ292468.1 | - | 5 | 396 | |||
AK030493.1 | - | 5 | 392 | |||
AK051870.1 | - | 5 | 413 | |||
BC056618.1 | - | 5 | 489 | |||
BC095940.1 | - | 5 | 316 | |||
BC145265.1 | - | 5 | 317 |