Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE2 SIRNA Assays ›
Gene Symbol:
SYNE2
Gene Name:
spectrin repeat containing nuclear envelope protein 2
Gene Aliases:
EDMD5, NUA, NUANCE, Nesp2, Nesprin-2, SYNE-2, TROPH
Chromosome Location:
Chr.14: 63761904 - 64226449 on Build GRCh38
UniGene ID:
Hs.745014
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23330
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015180.4
NP_055995.4
111
20339
NM_182910.2
NP_878914.1
5
1112
NM_182913.2
NP_878917.1
7
1217
NM_182914.2
NP_878918.2
112
20408
XM_005267454.1
XP_005267511.1
112
20450
XM_005267456.1
XP_005267513.1
112
20450
XM_005267457.1
XP_005267514.1
112
20408
XM_005267458.1
XP_005267515.1
111
20381
XM_005267459.1
XP_005267516.1
111
20339
XM_011536574.1
XP_011534876.1
112
20492
XM_011536575.2
XP_011534877.1
112
20469
XM_011536576.2
XP_011534878.1
113
20649
XM_011536577.2
XP_011534879.1
112
20416
XM_011536578.1
XP_011534880.1
112
20492
XM_011536579.1
XP_011534881.1
112
20450
XM_011536580.1
XP_011534882.1
112
20450
XM_011536581.1
XP_011534883.1
111
20423
XM_011536582.1
XP_011534884.1
111
20375
XM_017021101.1
XP_016876590.1
112
20494
XM_017021102.1
XP_016876591.1
111
20537
XM_017021103.1
XP_016876592.1
14
2529
XM_017021104.1
XP_016876593.1
14
2529
GenBank mRNA
AB023228.3
-
63
10080
AF357236.1
-
111
19878
AF435011.1
-
111
20321
AF495911.1
-
111
20306
AK001876.1
-
2
251
AK074055.1
-
23
3806
AK090430.1
-
29
4715
AK297874.1
-
7
1136
AK298465.1
-
6
850
AL080133.1
-
17
2852
AL117404.1
-
5
1634
AY061757.1
-
12
1943
AY061758.1
-
7
1214
AY061759.1
-
59
9642
AY184204.1
-
5
1634
AY184205.1
-
10
1824
BX537642.1
-
32
5153
BX538095.1
-
20
3315
BX640974.1
-
13
2154
CR749324.1
-
32
5313
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nuclear envelope organization
nuclear migration
establishment or maintenance of cell polarity
fibroblast migration
positive regulation of cell migration
nuclear migration along microfilament
protein localization to nucleus
centrosome localization
cytoskeletal anchoring at nuclear membrane
actin binding
protein binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0