Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GM14420 SIRNA Assays ›
          Gene Symbol:
          Gm14420 6230416C02Rik Etohi1 Gm14288 Gm14327 Gm14431 Gm14436 Gm14440 Gm8898 Gm8923
          Gene Name:
          predicted gene 14420
          RIKEN cDNA 6230416C02 gene
          ethanol induced 1
          predicted gene 14288
          predicted gene 14327
          predicted gene 14431
          predicted gene 14436
          predicted gene 14440
          predicted gene 8898
          predicted gene 8923
          Gene Aliases:
          OTTMUSG00000016588
          -
          -
          OTTMUSG00000016199
          OTTMUSG00000016328
          OTTMUSG00000016608
          OTTMUSG00000016614
          OTTMUSG00000016618
          OTTMUSG00000016203
          668009
          Chromosome Location:
          Chr.2: 177464742 - 177479194 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.2: 178023284 - 178035859 on Build GRCm38
          Chromosome Location:
          Chr.2: 175792312 - 175801038 on Build GRCm38
          Chromosome Location:
          Chr.2: 177897163 - 177927850 on Build GRCm38
          Chromosome Location:
          Chr.2: 176521004 - 176533821 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.2: 175275125 - 175286805 on Build GRCm38
          Chromosome Location:
          Chr.2: 175918283 - 175931100 on Build GRCm38
          Chromosome Location:
          Chr.2: 175470025 - 175480553 on Build GRCm38
          UniGene ID:
          Mm.460436
          Mm.484023
          Mm.422604
          -
          Mm.440737
          Mm.484983
          Mm.484023
          Mm.455970
          Mm.422667
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s233600

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gm14420
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177568.1 NP_001171039.1 2 204
          GenBank mRNA BC124559.1 - 2 204 30730885
          Gene Symbol: 6230416C02Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK018090.1 - 2 236
          BC056163.1 - 2 171
          Gene Symbol: Etohi1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177399.1 NP_001170870.1 2 195
          NM_001177400.1 NP_001170871.1 2 195
          GenBank mRNA AK005093.1 - 2 196
          Gene Symbol: Gm14288
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001033123.3 NP_001028295.2 2 177
          GenBank mRNA BC086907.1 - 2 177
          Gene Symbol: Gm14327
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC099593.1 - 2 255
          Gene Symbol: Gm14431
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177406.1 NP_001170877.1 2 209
          Gene Symbol: Gm14436
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242943.1 NP_001229872.1 160
          Gene Symbol: Gm14440
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199308.1 NP_001186237.1 2 176
          Gene Symbol: Gm8898
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177407.1 NP_001170878.1 2 209
          Gene Symbol: Gm8923
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242944.1 NP_001229873.1 2 195

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-gvwhr:80/100.66.78.51:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0