Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Sp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005343.2 | NP_001005343.1 | 2 | 2131 | ||
GenBank mRNA | AK135123.1 | - | 2 | 2130 | ||
AY591908.1 | - | 2 | 2143 |