Search
Search

Col2a1| Gene Symbol: Col2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113515.2 | NP_001106987.2 | 3, 4 | 362 | ||
| NM_031163.3 | NP_112440.2 | 4, 5 | 566 | |||
| GenBank mRNA | AK132311.1 | - | 3, 4 | 283 | ||
| BC051383.1 | - | 3, 4 | 273 | 6312433 | ||
| BC052326.1 | - | 3, 4 | 252 | 6315379 | ||
| BC082331.1 | - | 4, 5 | 499 | 30537724 | ||