Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Phip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081216.1 | NP_001074685.1 | 17 | 1912 | ||
GenBank mRNA | AB049460.1 | - | 17 | 1831 | ||
AJ303103.1 | - | 17 | 1808 | |||
AK032797.1 | - | 17 | 1951 | |||
AK049736.1 | - | 17 | 1848 | |||
AK050025.1 | - | 17 | 1858 | |||
AK136530.1 | - | 17 | 1850 | |||
BC009146.1 | - | 8 | 786 | |||
BC037036.1 | - | 17 | 1841 | |||
BC068022.1 | - | 9 | 903 |