Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H3I SIRNA Assays ›
          Gene Symbol:
          Hist1h3i Hist1h3a Hist1h3b Hist1h3c Hist1h3d Hist1h3e Hist1h3f Hist1h3g Hist1h3h
          Gene Name:
          histone cluster 1, H3i
          histone cluster 1, H3a
          histone cluster 1, H3b
          histone cluster 1, H3c
          histone cluster 1, H3d
          histone cluster 1, H3e
          histone cluster 1, H3f
          histone cluster 1, H3g
          histone cluster 1, H3h
          Gene Aliases:
          H3-H
          -
          H3-53
          H3-143
          H3-b
          H3-f, H3.1
          H3.2-221, Hist1
          H3.1-221, M32460
          H3-291
          Chromosome Location:
          Chr.13: 21782915 - 21783397 on Build GRCm38
          Chromosome Location:
          Chr.13: 23761885 - 23762386 on Build GRCm38
          Chromosome Location:
          Chr.13: 23752380 - 23752840 on Build GRCm38
          Chromosome Location:
          Chr.13: 23745042 - 23745521 on Build GRCm38
          Chromosome Location:
          Chr.13: 23575763 - 23576266 on Build GRCm38
          Chromosome Location:
          Chr.13: 23561896 - 23562365 on Build GRCm38
          Chromosome Location:
          Chr.13: 23544052 - 23544954 on Build GRCm38
          Chromosome Location:
          Chr.13: 23535418 - 23535909 on Build GRCm38
          Chromosome Location:
          Chr.13: 21717628 - 21718115 on Build GRCm38
          UniGene ID:
          Mm.390558
          Mm.221301
          -
          Mm.262517
          Mm.347699
          Mm.261658
          Mm.221389
          Mm.261657
          Mm.377874
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234218

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h3i
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178207.2 NP_835514.1 1 385
          GenBank mRNA AK155722.1 - 386
          BC107287.2 - 1 354
          BC116383.2 - 1 358 40101629
          BC127148.1 - 1 357
          Gene Symbol: Hist1h3a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013550.5 NP_038578.2 1 391
          GenBank mRNA AK006742.1 - 1 371
          AK018952.1 - 1 392
          BC115816.1 - 1 388 40105082
          Gene Symbol: Hist1h3b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178203.2 NP_835510.1 1 358
          Gene Symbol: Hist1h3c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175653.2 NP_783584.1 1 382
          GenBank mRNA BC120800.1 - 1 463 40129723
          BC120802.1 - 1 463 40129725
          Gene Symbol: Hist1h3d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178204.2 NP_835511.1 1 397
          GenBank mRNA Z30939.1 - 1 124
          Gene Symbol: Hist1h3e
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178205.2 NP_835512.1 1 358
          GenBank mRNA BC127042.1 - 1 351
          Gene Symbol: Hist1h3f
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013548.4 NP_038576.1 1 791
          GenBank mRNA AK010121.1 - 1 380
          BC101954.1 - 1 791 40044770
          BC101955.1 - 1 791 40044769
          BC101956.1 - 1 791 40044765
          BC103549.1 - 1 791 40044771
          Gene Symbol: Hist1h3g
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_145073.2 NP_659539.1 1 386
          GenBank mRNA BC125355.1 - 1 382 40129870
          BC125357.1 - 1 382 40129872
          Gene Symbol: Hist1h3h
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178206.2 NP_835513.1 1 389
          GenBank mRNA BC107285.2 - 1 356

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          nucleosome assembly
          DNA replication-dependent nucleosome assembly
          protein heterotetramerization
          regulation of gene silencing
          histone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0