Search
Search

Nanog| Gene Symbol: Nanog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289828.1 | NP_001276757.1 | 4 | 1091 | ||
| NM_001289830.1 | NP_001276759.1 | 5 | 891 | |||
| NM_001289831.1 | NP_001276760.1 | 5 | 875 | |||
| NM_028016.3 | NP_082292.1 | 4 | 1094 | |||
| GenBank mRNA | AB093574.1 | - | 4 | 1065 | ||
| AB126939.1 | - | 4 | 876 | |||
| AF507043.1 | - | 4 | 1066 | |||
| AK010332.1 | - | 4 | 1066 | |||
| AK167259.1 | - | 4 | 1066 | |||
| AY278951.1 | - | 4 | 943 | |||
| AY455282.1 | - | 5 | 875 | |||
| AY455285.1 | - | 5 | 891 | |||
| BC132017.1 | - | 4 | 1091 | 40130294 | ||
| BC137873.1 | - | 4 | 1091 | 8860892 | ||
| BC144995.1 | - | 4 | 1088 | 9053543 | ||
| BC144996.1 | - | 4 | 1091 | 9053544 | ||