Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MUP10 SIRNA Assays ›
          Gene Symbol:
          Mup10 Gm2083 LOC100048884 Mup1 Mup11 Mup12 Mup13 Mup14 Mup15 Mup16 Mup17 Mup19 Mup2 Mup7 Mup8 Mup9
          Gene Name:
          major urinary protein 10
          major urinary protein LOC100048885
          novel member of the major urinary protein (Mup) gene family
          major urinary protein 1
          major urinary protein 11
          major urinary protein 12
          major urinary protein 13
          major urinary protein 14
          major urinary protein 15
          major urinary protein 16
          major urinary protein 17
          major urinary protein 19
          major urinary protein 2
          major urinary protein 7
          major urinary protein 8
          major urinary protein 9
          Gene Aliases:
          2610016E04Rik, Mup8
          -
          Mup6
          Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
          Gm12549, OTTMUSG00000007431
          Gm2024
          Gm13513, Mup11, OTTMUSG00000012492
          Gm13514, Mup12, OTTMUSG00000012493
          Gm2068, Mup13
          -
          Gm12557, MUP 17, OTTMUSG00000007480
          100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
          AA589603, Mup-2, Mup4
          Gm12546, Mup3, OTTMUSG00000007428
          Gm12809, Mup5, OTTMUSG00000008509
          Gm14076, Mup2, Mup6, OTTMUSG00000015595
          Chromosome Location:
          Chr.4: 60578260 - 60582155 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.4: 61670177 - 61674098 on Build GRCm38
          Chromosome Location:
          Chr.4: 60498012 - 60501960 on Build GRCm38
          Chromosome Location:
          Chr.4: 60658466 - 60662395 on Build GRCm38
          Chromosome Location:
          Chr.4: 60737381 - 60741281 on Build GRCm38
          Chromosome Location:
          Chr.4: 61224307 - 61228286 on Build GRCm38
          Chromosome Location:
          Chr.4: 61300037 - 61303998 on Build GRCm38
          Chromosome Location:
          Chr.4: 61435787 - 61439714 on Build GRCm38
          Chromosome Location:
          Chr.4: 61515592 - 61519503 on Build GRCm38
          Chromosome Location:
          Chr.4: 61591929 - 61595853 on Build GRCm38
          Chromosome Location:
          Chr.4: 61778324 - 61782236 on Build GRCm38
          Chromosome Location:
          Chr.4: 60135913 - 60139872 on Build GRCm38
          Chromosome Location:
          Chr.4: 60066469 - 60070481 on Build GRCm38
          Chromosome Location:
          Chr.4: 60218620 - 60222599 on Build GRCm38
          Chromosome Location:
          Chr.4: 60418046 - 60421961 on Build GRCm38
          UniGene ID:
          Mm.335875
          Mm.422695
          Mm.335875
          Mm.335875
          Mm.482557
          Mm.482236
          Mm.443549
          Mm.458441
          Mm.237772
          Mm.422695
          Mm.475721
          Mm.457980
          Mm.335875
          Mm.457982
          Mm.457982
          Mm.482231
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234322

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mup10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122647.1 NP_001116119.1 4 411
          GenBank mRNA AJ309921.1 - 344
          AK011413.1 - 2 463
          CB948918.1 - 320
          EU882231.1 - 4 344
          Gene Symbol: Gm2083
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134644.1 NP_001128116.1 4 409
          Gene Symbol: LOC100048884
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199333.1 NP_001186262.1 4 416
          GenBank mRNA BC055353.1 - 391
          Gene Symbol: Mup1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163010.1 NP_001156482.1 4 455
          NM_001163011.1 NP_001156483.1 4 455
          NM_031188.2 NP_112465.2 4 455
          GenBank mRNA BC012221.1 - 4 401 4195753
          BC013649.1 - 389 4195401
          BC037152.1 - 279 4161709
          BC091744.1 - 392 5102271
          BC091775.1 - 4 402 5137341
          BC092254.1 - 389 5100458
          BC093515.1 - 389
          BC099597.1 - 392 947916
          BC100586.1 - 409 6769126
          BC106092.1 - 4 392 5098340
          M16355.1 - 4 455
          M27609.1 - 257
          M28649.1 - 402
          X03524.1 - 257
          X04115.1 - 213
          Gene Symbol: Mup11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164526.1 NP_001157998.1 442
          GenBank mRNA AK013972.1 - 4 419
          Gene Symbol: Mup12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199995.1 NP_001186924.1 4 410
          Gene Symbol: Mup13
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134674.1 NP_001128146.1 4 470
          Gene Symbol: Mup14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199999.1 NP_001186928.1 4 453
          Gene Symbol: Mup15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001200004.1 NP_001186933.1 4 416
          Gene Symbol: Mup16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199936.1 NP_001186865.1 4 410
          Gene Symbol: Mup17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001200006.1 NP_001186935.1 4 416
          Gene Symbol: Mup19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135127.2 NP_001128599.1 4 410
          GenBank mRNA AK013259.1 - 4 410
          EU882232.1 - 4 338
          Gene Symbol: Mup2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001045550.2 NP_001039015.1 4 414
          NM_001286096.1 NP_001273025.1 3 280
          NM_008647.4 NP_032673.3 4 414
          GenBank mRNA BC012259.1 - 403 4196403
          BC059097.1 - 389 4159984
          CB593503.1 - 412
          M16356.1 - 402
          Gene Symbol: Mup7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134675.1 NP_001128147.1 4 519
          GenBank mRNA EU882230.1 - 4 344
          Gene Symbol: Mup8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134676.1 NP_001128148.1 4 474
          Gene Symbol: Mup9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001281979.1 NP_001268908.1 4 410
          GenBank mRNA BI219655.1 - 4 391
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BC094452.1 - 390 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          transport
          molecular_function
          pheromone activity
          pheromone binding
          transporter activity
          small molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-4p4qf:80/100.66.72.111:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0