Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MUP10 SIRNA Assays ›
          Gene Symbol:
          Mup10 LOC100048884 Mup1 Mup11 Mup12 Mup14 Mup15 Mup16 Mup19 Mup2 Mup8 Mup9
          Gene Name:
          major urinary protein 10
          novel member of the major urinary protein (Mup) gene family
          major urinary protein 1
          major urinary protein 11
          major urinary protein 12
          major urinary protein 14
          major urinary protein 15
          major urinary protein 16
          major urinary protein 19
          major urinary protein 2
          major urinary protein 8
          major urinary protein 9
          Gene Aliases:
          2610016E04Rik, Mup8
          Mup6
          Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
          Gm12549, OTTMUSG00000007431
          Gm2024
          Gm13514, Mup12, OTTMUSG00000012493
          Gm2068, Mup13
          -
          100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
          AA589603, Mup-2, Mup4
          Gm12809, Mup5, OTTMUSG00000008509
          Gm14076, Mup2, Mup6, OTTMUSG00000015595
          Chromosome Location:
          Chr.4: 60578260 - 60582155 on Build GRCm38
          Chromosome Location:
          Chr.4: 61670177 - 61674098 on Build GRCm38
          Chromosome Location:
          Chr.4: 60498012 - 60501960 on Build GRCm38
          Chromosome Location:
          Chr.4: 60658466 - 60662395 on Build GRCm38
          Chromosome Location:
          Chr.4: 60737381 - 60741281 on Build GRCm38
          Chromosome Location:
          Chr.4: 61300037 - 61303998 on Build GRCm38
          Chromosome Location:
          Chr.4: 61435787 - 61439714 on Build GRCm38
          Chromosome Location:
          Chr.4: 61515592 - 61519503 on Build GRCm38
          Chromosome Location:
          Chr.4: 61778324 - 61782236 on Build GRCm38
          Chromosome Location:
          Chr.4: 60135913 - 60139872 on Build GRCm38
          Chromosome Location:
          Chr.4: 60218620 - 60222599 on Build GRCm38
          Chromosome Location:
          Chr.4: 60418046 - 60421961 on Build GRCm38
          UniGene ID:
          Mm.335875
          Mm.335875
          Mm.335875
          Mm.482557
          Mm.482236
          Mm.458441
          Mm.237772
          Mm.422695
          Mm.457980
          Mm.335875
          Mm.457982
          Mm.482231
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234323

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mup10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122647.1 NP_001116119.1 2 215
          GenBank mRNA AJ309921.1 - 148
          CB948918.1 - 124
          EU882231.1 - 2 148
          Gene Symbol: LOC100048884
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199333.1 NP_001186262.1 2 220
          GenBank mRNA BC055353.1 - 195
          Gene Symbol: Mup1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163010.1 NP_001156482.1 2 259
          NM_001163011.1 NP_001156483.1 2 259
          NM_031188.2 NP_112465.2 2 259
          GenBank mRNA BC012221.1 - 2 205 4195753
          BC013649.1 - 193 4195401
          BC091744.1 - 196 5102271
          BC091775.1 - 2 206 5137341
          BC092254.1 - 193 5100458
          BC093515.1 - 193
          BC099597.1 - 196 947916
          BC106092.1 - 2 196 5098340
          M16355.1 - 2 259
          M27609.1 - 61
          M28649.1 - 206
          X03524.1 - 61
          X04115.1 - 17
          Gene Symbol: Mup11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164526.1 NP_001157998.1 246
          GenBank mRNA AK013972.1 - 2 223
          Gene Symbol: Mup12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199995.1 NP_001186924.1 2 214
          Gene Symbol: Mup14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199999.1 NP_001186928.1 2 257
          Gene Symbol: Mup15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001200004.1 NP_001186933.1 2 220
          Gene Symbol: Mup16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199936.1 NP_001186865.1 2 214
          Gene Symbol: Mup19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135127.2 NP_001128599.1 2 214
          GenBank mRNA AK013259.1 - 2 214
          EU882232.1 - 2 142
          Gene Symbol: Mup2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001045550.2 NP_001039015.1 2 218
          NM_008647.4 NP_032673.3 2 218
          GenBank mRNA BC012259.1 - 207 4196403
          BC059097.1 - 193 4159984
          CB593503.1 - 216
          M16356.1 - 206
          Gene Symbol: Mup8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134676.1 NP_001128148.1 2 278
          Gene Symbol: Mup9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001281979.1 NP_001268908.1 2 214
          GenBank mRNA BI219655.1 - 2 195
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BC094452.1 - 194 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          pheromone activity
          pheromone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jww4n:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0