Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MUP19 SIRNA Assays ›
          Gene Symbol:
          Mup19 Gm2083 LOC100048884 Mup1 Mup10 Mup11 Mup12 Mup13 Mup14 Mup15 Mup16 Mup2 Mup7 Mup8 Mup9
          Gene Name:
          major urinary protein 19
          major urinary protein LOC100048885
          novel member of the major urinary protein (Mup) gene family
          major urinary protein 1
          major urinary protein 10
          major urinary protein 11
          major urinary protein 12
          major urinary protein 13
          major urinary protein 14
          major urinary protein 15
          major urinary protein 16
          major urinary protein 2
          major urinary protein 7
          major urinary protein 8
          major urinary protein 9
          Gene Aliases:
          100039247, Gm12552, Mup11, Mup14, Mup17, Mup8, OTTMUSG00000007472
          -
          Mup6
          Ltn-1, Lvtn-1, Mup-1, Mup-a, Mup10, Mup7, Up-1
          2610016E04Rik, Mup8
          Gm12549, OTTMUSG00000007431
          Gm2024
          Gm13513, Mup11, OTTMUSG00000012492
          Gm13514, Mup12, OTTMUSG00000012493
          Gm2068, Mup13
          -
          AA589603, Mup-2, Mup4
          Gm12546, Mup3, OTTMUSG00000007428
          Gm12809, Mup5, OTTMUSG00000008509
          Gm14076, Mup2, Mup6, OTTMUSG00000015595
          Chromosome Location:
          Chr.4: 61778324 - 61782236 on Build GRCm38
          Chromosome Location:
          -
          Chromosome Location:
          Chr.4: 61670177 - 61674098 on Build GRCm38
          Chromosome Location:
          Chr.4: 60498012 - 60501960 on Build GRCm38
          Chromosome Location:
          Chr.4: 60578260 - 60582155 on Build GRCm38
          Chromosome Location:
          Chr.4: 60658466 - 60662395 on Build GRCm38
          Chromosome Location:
          Chr.4: 60737381 - 60741281 on Build GRCm38
          Chromosome Location:
          Chr.4: 61224307 - 61228286 on Build GRCm38
          Chromosome Location:
          Chr.4: 61300037 - 61303998 on Build GRCm38
          Chromosome Location:
          Chr.4: 61435787 - 61439714 on Build GRCm38
          Chromosome Location:
          Chr.4: 61515592 - 61519503 on Build GRCm38
          Chromosome Location:
          Chr.4: 60135913 - 60139872 on Build GRCm38
          Chromosome Location:
          Chr.4: 60066469 - 60070481 on Build GRCm38
          Chromosome Location:
          Chr.4: 60218620 - 60222599 on Build GRCm38
          Chromosome Location:
          Chr.4: 60418046 - 60421961 on Build GRCm38
          UniGene ID:
          Mm.457980
          Mm.422695
          Mm.335875
          Mm.335875
          Mm.335875
          Mm.482557
          Mm.482236
          Mm.443549
          Mm.458441
          Mm.237772
          Mm.422695
          Mm.335875
          Mm.457982
          Mm.457982
          Mm.482231
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234412

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mup19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135127.2 NP_001128599.1 4 435
          GenBank mRNA AK013259.1 - 4 435
          EU882232.1 - 4 363
          Gene Symbol: Gm2083
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134644.1 NP_001128116.1 4 434
          Gene Symbol: LOC100048884
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199333.1 NP_001186262.1 4 441
          GenBank mRNA BC055353.1 - 416
          Gene Symbol: Mup1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163010.1 NP_001156482.1 4 480
          NM_001163011.1 NP_001156483.1 4 480
          NM_031188.2 NP_112465.2 4 480
          GenBank mRNA BC012221.1 - 4 426 4195753
          BC013649.1 - 414 4195401
          BC037152.1 - 304 4161709
          BC091744.1 - 417 5102271
          BC091775.1 - 4 427 5137341
          BC092254.1 - 414 5100458
          BC093515.1 - 414
          BC099597.1 - 417 947916
          BC100586.1 - 434 6769126
          BC106092.1 - 4 417 5098340
          M16355.1 - 4 480
          M27609.1 - 282
          M28649.1 - 427
          X03524.1 - 282
          X04115.1 - 238
          Gene Symbol: Mup10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001122647.1 NP_001116119.1 4 436
          GenBank mRNA AJ309921.1 - 369
          AK011413.1 - 2 488
          CB948918.1 - 345
          EU882231.1 - 4 369
          Gene Symbol: Mup11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001164526.1 NP_001157998.1 467
          GenBank mRNA AK013972.1 - 4 444
          Gene Symbol: Mup12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199995.1 NP_001186924.1 4 435
          Gene Symbol: Mup13
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134674.1 NP_001128146.1 4 495
          Gene Symbol: Mup14
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199999.1 NP_001186928.1 4 478
          Gene Symbol: Mup15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001200004.1 NP_001186933.1 4 441
          Gene Symbol: Mup16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001199936.1 NP_001186865.1 4 435
          Gene Symbol: Mup2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001045550.2 NP_001039015.1 4 439
          NM_001286096.1 NP_001273025.1 3 305
          NM_008647.4 NP_032673.3 4 439
          GenBank mRNA BC012259.1 - 428 4196403
          BC059097.1 - 414 4159984
          CB593503.1 - 437
          M16356.1 - 427
          Gene Symbol: Mup7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134675.1 NP_001128147.1 4 544
          GenBank mRNA EU882230.1 - 4 369
          Gene Symbol: Mup8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134676.1 NP_001128148.1 4 499
          Gene Symbol: Mup9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001281979.1 NP_001268908.1 4 435
          GenBank mRNA BI219655.1 - 4 416
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BC094452.1 - 415 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          pheromone activity
          pheromone binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-rm5rx:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0