Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gm5935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081657.1 | NP_001075126.1 | 8 | 636 | ||
GenBank mRNA | BC100411.1 | - | 8 | 636 | 30916114 |
Gene Symbol: Gm10058 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109969.2 | NP_001103439.1 | 8 | 636 | ||
GenBank mRNA | BU936093.1 | - | 639 |
Gene Symbol: Gm10096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102678.2 | NP_001096148.1 | 8 | 667 |
Gene Symbol: Gm10147 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099919.2 | NP_001093389.1 | 639 |
Gene Symbol: Gm10230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099347.2 | NP_001092817.1 | 8 | 636 |
Gene Symbol: Gm10486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109970.1 | NP_001103440.1 | 8 | 667 | ||
GenBank mRNA | BC087930.1 | - | 636 | 6773503 | ||
BC099540.1 | - | 8 | 607 | 6774523 | ||
X72697.1 | - | 654 |
Gene Symbol: Gm10487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100609.1 | NP_001094079.1 | 8 | 667 |
Gene Symbol: Gm10488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099325.2 | NP_001092795.1 | 8 | 636 | ||
GenBank mRNA | BC099541.1 | - | 8 | 636 | 30916030 |
Gene Symbol: Gm14525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162364.2 | NP_001155836.1 | 8 | 636 | ||
GenBank mRNA | BC048742.1 | - | 8 | 636 |
Gene Symbol: Gm14632 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100610.2 | NP_001094080.1 | 8 | 636 |
Gene Symbol: Gm14819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110250.2 | NP_001103720.1 | 636 |
Gene Symbol: Gm1993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102677.2 | NP_001096147.1 | 8 | 667 |
Gene Symbol: Gm2030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100445.2 | NP_001093915.1 | 8 | 667 |
Gene Symbol: Gm4297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100446.2 | NP_001093916.1 | 7 | 527 |
Gene Symbol: Gm4836 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009529.3 | NP_033555.1 | 8 | 667 | ||
GenBank mRNA | BC061169.1 | - | 8 | 667 | 6774630 |
Gene Symbol: Gm5168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025607.3 | NP_001020778.1 | 8 | 586 | ||
GenBank mRNA | BC099542.1 | - | 8 | 586 | 30917040 |
Gene Symbol: Gm5934 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100444.2 | NP_001093914.1 | 7 | 530 |
Gene Symbol: Gm6121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114754.1 | NP_001108226.1 | 8 | 636 |