Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GNAS SIRNA Assays ›
          Gene Symbol:
          Gnas
          Gene Name:
          GNAS (guanine nucleotide binding protein, alpha stimulating) complex locus
          Gene Aliases:
          5530400H20Rik, A930027G11Rik, C130027O20Rik, GPSA, GSP, Galphas, Gnas1, Gnasxl, Gsa, Nesp, Nespl, Oed-Sml, Oedsml, P1, P2, P3, PHP1A, PHP1B, POH
          Chromosome Location:
          Chr.2: 174283444 - 174346744 on Build GRCm38
          UniGene ID:
          Mm.125770
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s234484

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gnas
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_201617.1 NP_963911.1 2 2669
          GenBank mRNA AK030489.1 - 2 1600
          AK047990.1 - 1 1820
          AY519503.1 - 2 2669

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          tissue homeostasis
          positive regulation of protein phosphorylation
          endochondral ossification
          energy reserve metabolic process
          DNA methylation
          regulation of transcription from RNA polymerase II promoter
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          adenylate cyclase-activating dopamine receptor signaling pathway
          sensory perception of chemical stimulus
          post-embryonic development
          regulation of signal transduction
          positive regulation of sodium ion transport
          positive regulation of cAMP biosynthetic process
          embryonic hindlimb morphogenesis
          multicellular organism growth
          negative regulation of renal sodium excretion
          negative regulation of multicellular organism growth
          post-embryonic body morphogenesis
          response to drug
          positive regulation of GTPase activity
          skin development
          positive regulation of cAMP-mediated signaling
          positive regulation of osteoblast differentiation
          positive regulation of osteoclast differentiation
          negative regulation of blood pressure
          developmental growth
          embryonic cranial skeleton morphogenesis
          cognition
          cartilage development
          calcium ion homeostasis
          bone development
          hair follicle placode formation
          platelet aggregation
          response to parathyroid hormone
          cellular response to prostaglandin E stimulus
          genetic imprinting
          cellular response to catecholamine stimulus
          adenylate cyclase-activating adrenergic receptor signaling pathway
          nucleotide binding
          G-protein alpha-subunit binding
          GTPase activity
          signal transducer activity
          insulin-like growth factor receptor binding
          protein binding
          GTP binding
          guanyl nucleotide binding
          protein domain specific binding
          G-protein beta-subunit binding
          G-protein beta/gamma-subunit complex binding
          beta-2 adrenergic receptor binding
          D1 dopamine receptor binding
          mu-type opioid receptor binding
          ionotropic glutamate receptor binding
          alpha-tubulin binding
          metal ion binding
          corticotropin-releasing hormone receptor 1 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pqldv:80/100.66.72.197:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0