Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HIST1H4J SIRNA Assays ›
          Gene Symbol:
          Hist1h4j Hist1h4a Hist1h4b Hist1h4c Hist1h4d Hist1h4f Hist1h4h Hist1h4i Hist1h4k Hist1h4m Hist1h4n Hist2h4
          Gene Name:
          histone cluster 1, H4j
          histone cluster 1, H4a
          histone cluster 1, H4b
          histone cluster 1, H4c
          histone cluster 1, H4d
          histone cluster 1, H4f
          histone cluster 1, H4h
          histone cluster 1, H4i
          histone cluster 1, H4k
          histone cluster 1, H4m
          histone cluster 1, H4n
          histone cluster 2, H4
          Gene Aliases:
          -
          -
          -
          -
          -
          -
          1700024H08Rik
          -
          -
          Gm11275, OTTMUSG00000000446
          -
          H4, X04652
          Chromosome Location:
          Chr.13: 21735096 - 21735407 on Build GRCm38
          Chromosome Location:
          Chr.13: 23760795 - 23761249 on Build GRCm38
          Chromosome Location:
          Chr.13: 23756937 - 23757386 on Build GRCm38
          Chromosome Location:
          Chr.13: 23698084 - 23698458 on Build GRCm38
          Chromosome Location:
          Chr.13: 23581602 - 23581969 on Build GRCm38
          Chromosome Location:
          Chr.13: 23551286 - 23551643 on Build GRCm38
          Chromosome Location:
          Chr.13: 23531044 - 23531478 on Build GRCm38
          Chromosome Location:
          Chr.13: 22040960 - 22041362 on Build GRCm38
          Chromosome Location:
          Chr.13: 21750145 - 21750553 on Build GRCm38
          Chromosome Location:
          Chr.13: 21811746 - 21812150 on Build GRCm38
          Chromosome Location:
          Chr.13: 21831793 - 21832158 on Build GRCm38
          Chromosome Location:
          Chr.3: 96262934 - 96263317 on Build GRCm38
          UniGene ID:
          Mm.486099
          Mm.228709
          Mm.261642
          Mm.261662
          Mm.260530
          Mm.442307
          Mm.158272
          Mm.14775
          Mm.255646
          Mm.442307
          Mm.261664
          Mm.227295
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s234605

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hist1h4j
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178210.1 NP_835582.1 1 181
          GenBank mRNA AK010085.1 - 1 217
          Gene Symbol: Hist1h4a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178192.2 NP_835499.1 1 273
          GenBank mRNA AK016310.1 - 1 255
          BC119241.1 - 1 273 8733990
          BC119243.1 - 1 273 8733992
          Gene Symbol: Hist1h4b
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178193.2 NP_835500.1 1 265
          GenBank mRNA BC132186.1 - 1 265 40130463
          BC132212.1 - 1 265 40130489
          Gene Symbol: Hist1h4c
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178208.2 NP_835515.1 1 190
          GenBank mRNA BC115450.1 - 1 181 40099776
          BC115451.1 - 1 174
          BC139809.1 - 1 181 40099779
          Gene Symbol: Hist1h4d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175654.2 NP_783585.1 1 181
          GenBank mRNA BC119611.2 - 1 180
          Gene Symbol: Hist1h4f
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175655.2 NP_783586.1 1 181
          GenBank mRNA BC115446.1 - 1 174
          Gene Symbol: Hist1h4h
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_153173.4 NP_694813.1 1 252
          GenBank mRNA AA028777.1 - 1 250
          AK007642.1 - 1 248
          BC028550.1 - 1 193 1398106
          BC058529.1 - 1 189 6817839
          BC087952.1 - 1 241 6787571
          Gene Symbol: Hist1h4i
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175656.3 NP_783587.1 1 220
          GenBank mRNA AK011560.1 - 1 221
          BC019757.1 - 1 210
          BC052219.1 - 1 193
          BC092144.1 - 1 210 30612926
          BC125598.1 - 1 204 40130113
          BC125600.1 - 1 204 40130115
          Gene Symbol: Hist1h4k
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_178211.2 NP_835583.1 1 229
          GenBank mRNA AK139521.1 - 1 216
          BC096767.1 - 196
          BC117010.1 - 1 229 40126329
          BC117012.1 - 1 229 40126331
          BC119612.1 - 1 181 40061668
          Gene Symbol: Hist1h4m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195421.1 NP_001182350.1 1 219
          GenBank mRNA BC139425.1 - 243 8862447
          BC139437.1 - 243 8862459
          Gene Symbol: Hist1h4n
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_175657.2 NP_783588.1 1 181
          GenBank mRNA BC115448.2 - 1 181 40099764
          BC152397.1 - 1 181 40099768
          Gene Symbol: Hist2h4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_033596.3 NP_291074.1 1 194
          GenBank mRNA BC051566.1 - 1 202
          BC057955.1 - 188 5359141
          BX520915.1 - 1 194

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_003548, NM_001034077, BC093969, BC108260, CR542172, and others.
          This siRNA also targets the following transcripts in Rat: NM_001271221, NM_022686, XM_008771773, XM_008771675, XM_008771676, and others.

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleosome assembly
          DNA replication-dependent nucleosome assembly
          DNA replication-independent nucleosome assembly
          negative regulation of megakaryocyte differentiation
          protein heterotetramerization
          histone binding
          poly(A) RNA binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0