Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RGD1564744 SIRNA Assays ›
          Gene Symbol:
          RGD1564744 Rplp1
          Gene Name:
          similar to 60S acidic ribosomal protein P1
          ribosomal protein, large, P1
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.7: 18613039 - 18613557 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 66862143 - 66863476 on Build Rnor_6.0
          UniGene ID:
          -
          Rn.973
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s234845

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RGD1564744
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006241133.2 XP_006241195.1 2 428
          XM_006241134.2 XP_006241196.1 2 422
          XM_006241135.2 XP_006241197.1 2 422
          XM_008765205.1 XP_008763427.1 2 428
          XM_234961.5 XP_234961.1 1 482
          Gene Symbol: Rplp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007604.2 NP_001007605.1 3 460
          GenBank mRNA BC058151.1 - 3 463 6888011
          FQ209966.1 - 3 462
          FQ210237.1 - 3 459
          FQ210375.1 - 3 459
          FQ210407.1 - 3 459
          FQ210632.1 - 3 461
          FQ210864.1 - 3 459
          FQ210988.1 - 3 462
          FQ211097.1 - 3 462
          FQ211327.1 - 3 460
          FQ211648.1 - 3 459
          FQ212351.1 - 3 459
          FQ214973.1 - 3 461
          FQ215065.1 - 3 459
          FQ216905.1 - 3 463
          FQ216921.1 - 3 462
          FQ216959.1 - 3 458
          FQ216979.1 - 3 462
          FQ217216.1 - 3 462
          FQ217313.1 - 3 460
          FQ220167.1 - 3 460
          FQ220212.1 - 3 461
          FQ220998.1 - 3 459
          FQ221027.1 - 3 460
          FQ221120.1 - 3 460
          FQ221137.1 - 3 459
          FQ221201.1 - 3 462
          FQ221359.1 - 3 460
          FQ221455.1 - 3 464
          FQ221567.1 - 3 462
          FQ221595.1 - 3 460
          FQ221620.1 - 3 459
          FQ221698.1 - 3 466
          FQ221717.1 - 3 459
          FQ221756.1 - 3 459
          FQ221784.1 - 3 459
          FQ221894.1 - 3 459
          FQ221969.1 - 3 459
          FQ221980.1 - 3 459
          FQ222031.1 - 3 459
          FQ222178.1 - 3 459
          FQ222307.1 - 3 461
          FQ222468.1 - 3 461
          FQ222559.1 - 3 459
          FQ222618.1 - 3 462
          FQ222784.1 - 3 459
          FQ222801.1 - 3 460
          FQ222805.1 - 3 459
          FQ222816.1 - 3 460
          FQ222923.1 - 3 460
          FQ222965.1 - 3 459
          FQ222966.1 - 3 458
          FQ223013.1 - 3 460
          FQ223016.1 - 3 461
          FQ223066.1 - 3 461
          FQ223337.1 - 3 459
          FQ223344.1 - 3 460
          FQ223388.1 - 3 459
          FQ223390.1 - 3 460
          FQ223470.1 - 3 462
          FQ223512.1 - 3 460
          FQ223537.1 - 3 460
          FQ223567.1 - 3 459
          FQ223591.1 - 3 459
          FQ223674.1 - 3 441
          FQ223769.1 - 3 460
          FQ224482.1 - 3 429
          FQ224625.1 - 3 462
          FQ224766.1 - 3 460
          FQ228323.1 - 3 459
          FQ228361.1 - 3 459
          FQ228390.1 - 3 460
          FQ228490.1 - 3 460
          FQ228528.1 - 3 456
          FQ228539.1 - 3 459
          FQ228605.1 - 3 463
          FQ228650.1 - 3 459
          FQ228990.1 - 3 466
          FQ229009.1 - 3 459
          FQ229050.1 - 3 459
          FQ229318.1 - 3 460
          FQ230089.1 - 3 460
          X15097.1 - 3 430

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_018853, U29402, AK021242, BC157910, BC058685, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0