Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP1 SIRNA Assays ›
          Gene Symbol:
          Rplp1 RGD1564744
          Gene Name:
          ribosomal protein, large, P1
          similar to 60S acidic ribosomal protein P1
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.8: 66862143 - 66863476 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 18613039 - 18613557 on Build Rnor_6.0
          UniGene ID:
          Rn.973
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s234846

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007604.2 NP_001007605.1 3 458
          GenBank mRNA BC058151.1 - 3 461 6888011
          FQ209966.1 - 3 460
          FQ210237.1 - 3 457
          FQ210375.1 - 3 457
          FQ210407.1 - 3 457
          FQ210632.1 - 3 459
          FQ210864.1 - 3 457
          FQ210988.1 - 3 460
          FQ211097.1 - 3 460
          FQ211327.1 - 3 458
          FQ211648.1 - 3 457
          FQ212351.1 - 3 457
          FQ214973.1 - 3 459
          FQ215065.1 - 3 457
          FQ216905.1 - 3 461
          FQ216921.1 - 3 460
          FQ216959.1 - 3 456
          FQ216979.1 - 3 460
          FQ217216.1 - 3 460
          FQ217313.1 - 3 458
          FQ220167.1 - 3 458
          FQ220998.1 - 3 457
          FQ221027.1 - 3 458
          FQ221120.1 - 3 458
          FQ221137.1 - 3 457
          FQ221201.1 - 3 460
          FQ221359.1 - 3 458
          FQ221455.1 - 3 462
          FQ221567.1 - 3 460
          FQ221595.1 - 3 458
          FQ221620.1 - 3 457
          FQ221698.1 - 3 464
          FQ221717.1 - 3 457
          FQ221756.1 - 3 457
          FQ221784.1 - 3 457
          FQ221894.1 - 3 457
          FQ221969.1 - 3 457
          FQ221980.1 - 3 457
          FQ222031.1 - 3 457
          FQ222178.1 - 3 457
          FQ222307.1 - 3 459
          FQ222468.1 - 3 459
          FQ222559.1 - 3 457
          FQ222608.1 - 3 460
          FQ222618.1 - 3 460
          FQ222784.1 - 3 457
          FQ222801.1 - 3 458
          FQ222805.1 - 3 457
          FQ222816.1 - 3 458
          FQ222923.1 - 3 458
          FQ222965.1 - 3 457
          FQ222966.1 - 3 456
          FQ223013.1 - 3 458
          FQ223016.1 - 3 459
          FQ223066.1 - 3 459
          FQ223337.1 - 3 457
          FQ223344.1 - 3 458
          FQ223388.1 - 3 457
          FQ223390.1 - 3 458
          FQ223470.1 - 3 460
          FQ223512.1 - 3 458
          FQ223537.1 - 3 458
          FQ223567.1 - 3 457
          FQ223591.1 - 3 457
          FQ223674.1 - 3 439
          FQ223769.1 - 3 458
          FQ224482.1 - 3 427
          FQ224625.1 - 3 460
          FQ224766.1 - 3 458
          FQ228323.1 - 3 457
          FQ228361.1 - 3 457
          FQ228390.1 - 3 458
          FQ228490.1 - 3 458
          FQ228528.1 - 3 454
          FQ228539.1 - 3 457
          FQ228605.1 - 3 461
          FQ228650.1 - 3 457
          FQ228990.1 - 3 464
          FQ229009.1 - 3 457
          FQ229050.1 - 3 457
          FQ229318.1 - 3 458
          FQ230089.1 - 3 458
          X15097.1 - 3 428
          Gene Symbol: RGD1564744
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006241133.2 XP_006241195.1 2 426
          XM_006241134.2 XP_006241196.1 2 420
          XM_006241135.2 XP_006241197.1 2 420
          XM_008765205.1 XP_008763427.1 2 426
          XM_234961.5 XP_234961.1 1 480

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_018853, U29402, AK021242, BC157910, BC058685, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0