Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP1 SIRNA Assays ›
          Gene Symbol:
          Rplp1 RGD1564744
          Gene Name:
          ribosomal protein, large, P1
          similar to 60S acidic ribosomal protein P1
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.8: 66862143 - 66863476 on Build Rnor_6.0
          Chromosome Location:
          Chr.7: 18613039 - 18613557 on Build Rnor_6.0
          UniGene ID:
          Rn.973
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s234847

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007604.2 NP_001007605.1 3 471
          GenBank mRNA BC058151.1 - 3 474 6888011
          FQ209966.1 - 3 473
          FQ210237.1 - 3 470
          FQ210375.1 - 3 470
          FQ210407.1 - 3 470
          FQ210632.1 - 3 472
          FQ210864.1 - 3 470
          FQ210988.1 - 3 473
          FQ211097.1 - 3 473
          FQ211327.1 - 3 471
          FQ211648.1 - 3 470
          FQ212351.1 - 3 470
          FQ214973.1 - 3 472
          FQ215065.1 - 3 470
          FQ216905.1 - 3 474
          FQ216921.1 - 3 473
          FQ216959.1 - 3 469
          FQ216979.1 - 3 473
          FQ217216.1 - 3 473
          FQ217313.1 - 3 471
          FQ220167.1 - 3 471
          FQ220212.1 - 3 472
          FQ220998.1 - 3 470
          FQ221027.1 - 3 471
          FQ221120.1 - 3 471
          FQ221137.1 - 3 470
          FQ221201.1 - 3 473
          FQ221359.1 - 3 471
          FQ221455.1 - 3 475
          FQ221567.1 - 3 473
          FQ221595.1 - 3 471
          FQ221620.1 - 3 470
          FQ221698.1 - 3 477
          FQ221717.1 - 3 470
          FQ221756.1 - 3 470
          FQ221784.1 - 3 470
          FQ221894.1 - 3 470
          FQ221969.1 - 3 470
          FQ221980.1 - 3 470
          FQ222031.1 - 3 470
          FQ222178.1 - 3 470
          FQ222307.1 - 3 472
          FQ222468.1 - 3 472
          FQ222559.1 - 3 470
          FQ222618.1 - 3 473
          FQ222801.1 - 3 471
          FQ222805.1 - 3 470
          FQ222816.1 - 3 471
          FQ222879.1 - 3 466
          FQ222923.1 - 3 471
          FQ222965.1 - 3 470
          FQ223013.1 - 3 471
          FQ223016.1 - 3 472
          FQ223066.1 - 3 472
          FQ223337.1 - 3 470
          FQ223344.1 - 3 471
          FQ223388.1 - 3 470
          FQ223390.1 - 3 471
          FQ223470.1 - 3 473
          FQ223512.1 - 3 471
          FQ223537.1 - 3 471
          FQ223567.1 - 3 470
          FQ223591.1 - 3 470
          FQ223674.1 - 3 452
          FQ223769.1 - 3 471
          FQ224482.1 - 3 440
          FQ224625.1 - 3 473
          FQ224766.1 - 3 471
          FQ228323.1 - 3 470
          FQ228361.1 - 3 470
          FQ228390.1 - 3 471
          FQ228490.1 - 3 471
          FQ228528.1 - 3 467
          FQ228539.1 - 3 470
          FQ228605.1 - 3 474
          FQ228650.1 - 3 470
          FQ228990.1 - 3 477
          FQ229009.1 - 3 470
          FQ229050.1 - 3 470
          FQ229318.1 - 3 471
          FQ230089.1 - 3 471
          X15097.1 - 3 441
          Gene Symbol: RGD1564744
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_006241133.2 XP_006241195.1 2 439
          XM_006241134.2 XP_006241196.1 2 433
          XM_006241135.2 XP_006241197.1 2 433
          XM_008765205.1 XP_008763427.1 2 439
          XM_234961.5 XP_234961.1 1 493

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          structural constituent of ribosome

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0