Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HBA2 SIRNA Assays ›
          Gene Symbol:
          Hba2 Hba1
          Gene Name:
          hemoglobin, alpha 2
          hemoglobin, alpha 1
          Gene Aliases:
          -
          HBAM, Hba-a1, Hba-a2
          Chromosome Location:
          Chr.10: 15589364 - 15590207 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 15602794 - 15603649 on Build Rnor_6.0
          UniGene ID:
          Rn.203003
          Rn.107334
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s234978

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hba2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007722.1 NP_001007723.1 133
          GenBank mRNA FQ209950.1 - 1, 2 131
          FQ210108.1 - 1, 2 131
          FQ210256.1 - 1, 2 131
          FQ210264.1 - 1, 2 131
          FQ210372.1 - 1, 2 136
          FQ210420.1 - 1, 2 131
          FQ210570.1 - 1, 2 131
          FQ210694.1 - 1, 2 131
          FQ211180.1 - 1, 2 131
          FQ211270.1 - 1, 2 131
          FQ211366.1 - 1, 2 131
          FQ211672.1 - 1, 2 131
          FQ211887.1 - 1, 2 131
          FQ213126.1 - 1, 2 131
          FQ214258.1 - 1, 2 131
          FQ215087.1 - 1, 2 131
          FQ217874.1 - 1, 2 131
          FQ218140.1 - 1, 2 133
          FQ218297.1 - 1, 2 137
          FQ221177.1 - 1, 2 131
          FQ221489.1 - 1, 2 131
          FQ222342.1 - 1, 2 131
          FQ222639.1 - 1, 2 130
          FQ222693.1 - 1, 2 129
          FQ223754.1 - 1, 2 133
          FQ223838.1 - 1, 2 157
          FQ224218.1 - 1, 2 140
          FQ224339.1 - 1, 2 133
          FQ224679.1 - 1, 2 131
          FQ225311.1 - 1, 2 131
          FQ225451.1 - 1, 2 131
          FQ225711.1 - 1, 2 131
          FQ225789.1 - 1, 2 133
          FQ226185.1 - 1, 2 131
          FQ226226.1 - 286
          FQ226238.1 - 1, 2 121
          FQ226556.1 - 1, 2 131
          FQ226574.1 - 1, 2 131
          FQ226576.1 - 1, 2 131
          FQ226594.1 - 1, 2 131
          FQ226619.1 - 1, 2 131
          FQ226919.1 - 1, 2 129
          FQ226927.1 - 1, 2 131
          FQ228017.1 - 1, 2 133
          FQ228292.1 - 1, 2 131
          FQ228298.1 - 1, 2 131
          FQ228306.1 - 1, 2 131
          FQ228588.1 - 1, 2 131
          FQ229556.1 - 1, 2 131
          FQ230499.1 - 1, 2 131
          FQ230501.1 - 1, 2 131
          FQ230536.1 - 1, 2 131
          FQ230819.1 - 1, 2 133
          FQ230868.1 - 1, 2 131
          FQ230877.1 - 1, 2 133
          FQ231222.1 - 1, 2 131
          FQ231239.1 - 1, 2 126
          FQ231246.1 - 1, 2 131
          FQ231250.1 - 1, 2 131
          FQ231268.1 - 1, 2 130
          FQ231572.1 - 1, 2 131
          FQ231902.1 - 1, 2 131
          FQ232213.1 - 1, 2 131
          FQ234805.1 - 1, 2 131
          FQ235117.1 - 1, 2 132
          Gene Symbol: Hba1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013096.1 NP_037228.1 1, 2 133
          GenBank mRNA BC059150.1 - 1, 2 132 6918464
          BC091567.1 - 120 7458326
          BC160818.1 - 1, 2 116 7375173
          FQ209733.1 - 1, 2 133
          FQ209744.1 - 1, 2 133
          FQ209909.1 - 1, 2 133
          FQ210001.1 - 1, 2 133
          FQ210150.1 - 1, 2 131
          FQ210186.1 - 1, 2 131
          FQ210277.1 - 1, 2 131
          FQ210349.1 - 1, 2 131
          FQ210635.1 - 1, 2 133
          FQ210643.1 - 1, 2 131
          FQ210690.1 - 1, 2 132
          FQ210723.1 - 1, 2 131
          FQ210862.1 - 1, 2 131
          FQ210866.1 - 1, 2 131
          FQ211077.1 - 1, 2 133
          FQ211217.1 - 1, 2 131
          FQ211220.1 - 1, 2 133
          FQ211232.1 - 1, 2 132
          FQ211239.1 - 1, 2 131
          FQ211374.1 - 1, 2 131
          FQ211431.1 - 1, 2 131
          FQ211513.1 - 1, 2 131
          FQ211520.1 - 1, 2 131
          FQ211755.1 - 1, 2 131
          FQ211911.1 - 1, 2 131
          FQ211972.1 - 1, 2 131
          FQ212025.1 - 1, 2 133
          FQ212144.1 - 1, 2 133
          FQ212153.1 - 1, 2 133
          FQ212186.1 - 1, 2 131
          FQ212232.1 - 1, 2 133
          FQ212457.1 - 1, 2 131
          FQ212854.1 - 1, 2 131
          FQ213301.1 - 1, 2 131
          FQ214066.1 - 1, 2 131
          FQ214223.1 - 1, 2 131
          FQ216243.1 - 1, 2 131
          FQ217723.1 - 1, 2 131
          FQ217724.1 - 1, 2 131
          FQ217869.1 - 1, 2 131
          FQ217988.1 - 1, 2 133
          FQ218117.1 - 1, 2 131
          FQ218146.1 - 131
          FQ218257.1 - 1, 2 187
          FQ218612.1 - 1, 2 130
          FQ218968.1 - 1, 2 133
          FQ219715.1 - 1, 2 131
          FQ220786.1 - 1, 2 131
          FQ221656.1 - 1, 2 132
          FQ221912.1 - 1, 2 133
          FQ221952.1 - 1, 2 131
          FQ222431.1 - 1, 2 131
          FQ222454.1 - 1, 2 131
          FQ222750.1 - 1, 2 133
          FQ223017.1 - 1, 2 132
          FQ224581.1 - 1, 2 131
          FQ225427.1 - 1, 2 132
          FQ225440.1 - 1, 2 132
          FQ225452.1 - 1, 2 131
          FQ225777.1 - 1, 2 131
          FQ225808.1 - 1, 2 134
          FQ226118.1 - 1, 2 133
          FQ226173.1 - 1, 2 133
          FQ226175.1 - 1, 2 131
          FQ226203.1 - 1, 2 131
          FQ226615.1 - 1, 2 131
          FQ227082.1 - 1, 2 131
          FQ228301.1 - 1, 2 131
          FQ228436.1 - 131
          FQ230483.1 - 1, 2 133
          FQ230901.1 - 1, 2 131
          FQ231201.1 - 1, 2 123
          FQ231290.1 - 1, 2 131
          FQ232272.1 - 1, 2 131
          FQ232481.1 - 1, 2 131
          FQ234683.1 - 1, 2 131
          M17083.1 - 1, 2 133

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of cell death
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          erythrocyte development
          protein heterooligomerization
          oxidation-reduction process
          oxygen transport
          negative regulation of blood pressure
          regulation of sensory perception of pain
          peroxidase activity
          haptoglobin binding
          beta-amyloid binding
          oxygen transporter activity
          iron ion binding
          protein binding
          oxygen binding
          heme binding
          organic acid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-njpwx:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0