Search
Search

Map7d1| Gene Symbol: Map7d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109438.1 | NP_001102908.1 | 11, 12 | 2063 | ||
| XM_006238901.2 | XP_006238963.1 | 10, 11 | 2139 | |||
| XM_006238902.2 | XP_006238964.1 | 10, 11 | 2130 | |||
| XM_006238903.2 | XP_006238965.1 | 9, 10 | 2028 | |||
| XM_006238904.2 | XP_006238966.1 | 9, 10 | 2019 | |||
| XM_006238906.2 | XP_006238968.1 | 10, 11 | 1923 | |||
| GenBank mRNA | FQ216284.1 | - | 1, 2 | 208 | ||