Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wbscr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109499.2 | NP_001102969.1 | 5 | 520 | ||
XM_006249190.2 | XP_006249252.1 | 6 | 606 | |||
XM_006249191.2 | XP_006249253.1 | 6 | 606 |