Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NEDD4L SIRNA Assays ›
          Gene Symbol:
          NEDD4L
          Gene Name:
          neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
          Gene Aliases:
          NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
          Chromosome Location:
          Chr.18: 58044362 - 58401540 on Build GRCh38
          UniGene ID:
          Hs.185677
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s23569

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NEDD4L
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144964.1 NP_001138436.1 23 2472
          NM_001144965.1 NP_001138437.1 23 2424
          NM_001144966.2 NP_001138438.1 22 2349
          NM_001144967.2 NP_001138439.1 23 2518
          NM_001144968.1 NP_001138440.1 23 2414
          NM_001144969.1 NP_001138441.1 22 2354
          NM_001144970.2 NP_001138442.1 21 2397
          NM_001144971.1 NP_001138443.1 23 2442
          NM_001243960.1 NP_001230889.1 21 2326
          NM_015277.5 NP_056092.2 22 2458
          XM_005266658.4 XP_005266715.2 20 2449
          XM_005266660.4 XP_005266717.2 19 2393
          XM_005266663.4 XP_005266720.2 18 2333
          XM_006722421.3 XP_006722484.2 19 2388
          XM_006722424.3 XP_006722487.2 16 1941
          XM_006722425.3 XP_006722488.2 15 1885
          XM_006722426.3 XP_006722489.1 25 2666
          XM_006722428.3 XP_006722491.1 24 2588
          XM_006722430.3 XP_006722493.1 23 2496
          XM_011525887.2 XP_011524189.1 25 2646
          XM_017025676.1 XP_016881165.1 17 2138
          XM_017025677.1 XP_016881166.1 16 2082
          XM_017025678.1 XP_016881167.1 23 2527
          XM_017025679.1 XP_016881168.1 22 2442
          XM_017025680.1 XP_016881169.1 19 2130
          XM_017025681.1 XP_016881170.1 18 1932
          GenBank mRNA AB007899.1 - 21 2297
          AB071179.1 - 22 2289
          AF210730.1 - 22 2249
          AF385931.1 - 23 2473
          AK294963.1 - 22 2349
          AK300444.1 - 22 2300
          AL137469.1 - 18 1770
          AY112983.1 - 23 2424
          AY112984.2 - 23 2414
          AY112985.2 - 22 2358
          AY312514.1 - 23 2235
          BC000621.2 - 17 1875
          BC019345.1 - 17 1875
          BC032597.1 - 21 2353 5528964
          BQ215431.1 - 2 82
          DQ181796.1 - 23 2442

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein polyubiquitination
          regulation of membrane depolarization
          protein monoubiquitination
          sodium ion transport
          cellular sodium ion homeostasis
          excretion
          response to metal ion
          protein ubiquitination
          viral life cycle
          water homeostasis
          ion transmembrane transport
          regulation of ion transmembrane transport
          regulation of protein catabolic process
          regulation of membrane potential
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of protein catabolic process
          positive regulation of endocytosis
          regulation of membrane repolarization
          protein K48-linked ubiquitination
          ventricular cardiac muscle cell action potential
          regulation of potassium ion transmembrane transporter activity
          negative regulation of potassium ion transmembrane transporter activity
          negative regulation of potassium ion transmembrane transport
          negative regulation of sodium ion transmembrane transport
          positive regulation of dendrite extension
          negative regulation of protein localization to cell surface
          negative regulation of sodium ion transmembrane transporter activity
          positive regulation of caveolin-mediated endocytosis
          ubiquitin-protein transferase activity
          protein binding
          potassium channel regulator activity
          ligase activity
          sodium channel regulator activity
          potassium channel inhibitor activity
          sodium channel inhibitor activity
          ion channel binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0