Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIF SIRNA Assays ›
          Gene Symbol:
          Mif LOC103694877
          Gene Name:
          macrophage migration inhibitory factor (glycosylation-inhibiting factor)
          macrophage migration inhibitory factor
          Gene Aliases:
          -
          -
          Chromosome Location:
          Chr.20: 13732114 - 13732979 on Build Rnor_6.0
          Chromosome Location:
          Chr.Un|NW_007906821: 11287 - 12316 on Build Rnor_6.0
          UniGene ID:
          Rn.2661
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s235781

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mif
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031051.1 NP_112313.1 2 222
          GenBank mRNA BC061545.1 - 2 206 6920605
          FQ210474.1 - 2 204
          FQ213295.1 - 2 191
          FQ217414.1 - 2 202
          FQ222835.1 - 2 218
          FQ223656.1 - 2 179
          FQ229019.1 - 2 202
          FQ229084.1 - 2 202
          S73424.1 - 2 187
          U20999.1 - 2 193
          U62326.1 - 2 222
          Gene Symbol: LOC103694877
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008773846.1 XP_008772068.1 2 386

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_010798, AK008939, AK010691, BC100605, L10613, and others.

          Panther Classification:

          Molecular Function -

          decarboxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          prostaglandin biosynthetic process
          positive regulation of protein phosphorylation
          hair follicle development
          brain renin-angiotensin system
          positive regulation of acute inflammatory response
          negative regulation of mature B cell apoptotic process
          inflammatory response
          cell surface receptor signaling pathway
          spermatogenesis
          brain development
          aging
          cell aging
          metabolic process
          cell proliferation
          response to heat
          response to mechanical stimulus
          response to hormone
          response to inorganic substance
          negative regulation of gene expression
          positive regulation of protein kinase A signaling
          positive regulation of smooth muscle cell migration
          carboxylic acid metabolic process
          DNA damage response, signal transduction by p53 class mediator
          positive regulation of B cell proliferation
          positive regulation of lipopolysaccharide-mediated signaling pathway
          negative regulation of cellular protein metabolic process
          response to estradiol
          response to lipopolysaccharide
          response to progesterone
          response to menaquinone
          response to insulin
          negative regulation of myeloid cell apoptotic process
          positive regulation of peptidyl-serine phosphorylation
          response to vitamin E
          wound healing
          regulation of cell proliferation
          positive regulation of phosphorylation
          response to drug
          drinking behavior
          negative regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of MAP kinase activity
          negative regulation of DNA damage response, signal transduction by p53 class mediator
          skin development
          innate immune response
          positive regulation of glycolytic process
          positive regulation of glucose import
          positive regulation of fibroblast proliferation
          positive regulation of axon regeneration
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of immune response
          positive chemotaxis
          response to glucocorticoid
          positive regulation of prostaglandin secretion involved in immune response
          positive regulation of myeloid leukocyte cytokine production involved in immune response
          protein homotrimerization
          cellular response to hydrogen peroxide
          positive regulation of ERK1 and ERK2 cascade
          negative regulation of cell cycle arrest
          cellular response to hypoxia
          positive regulation of arachidonic acid secretion
          negative regulation of cell aging
          negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          dopachrome isomerase activity
          receptor binding
          cytokine activity
          cytokine receptor binding
          protein complex binding
          chemoattractant activity
          phenylpyruvate tautomerase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0