Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23610
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
133
24439
NM_182961.3
NP_892006.3
134
24817
XM_006715407.1
XP_006715470.1
134
24730
XM_006715408.2
XP_006715471.1
134
24722
XM_006715409.1
XP_006715472.1
133
24709
XM_006715410.2
XP_006715473.1
134
24734
XM_006715411.1
XP_006715474.1
133
24679
XM_006715412.2
XP_006715475.1
134
24719
XM_006715413.2
XP_006715476.1
134
24734
XM_006715414.1
XP_006715477.1
132
24658
XM_006715415.2
XP_006715478.1
134
24734
XM_006715416.2
XP_006715479.1
134
24719
XM_006715417.2
XP_006715480.1
133
24593
XM_006715420.2
XP_006715483.1
133
24581
XM_006715421.2
XP_006715484.1
133
24578
XM_006715422.1
XP_006715485.1
133
24571
XM_006715423.2
XP_006715486.1
134
24734
XM_006715424.2
XP_006715487.1
134
24734
XM_006715425.2
XP_006715488.1
134
24734
XM_011535641.2
XP_011533943.1
134
24731
XM_011535642.2
XP_011533944.1
134
24719
XM_011535643.1
XP_011533945.1
133
24565
XM_011535644.1
XP_011533946.1
120
22781
XM_011535645.2
XP_011533947.1
116
22174
XM_017010608.1
XP_016866097.1
135
24849
XM_017010609.1
XP_016866098.1
134
24656
XM_017010610.1
XP_016866099.1
133
24635
XM_017010611.1
XP_016866100.1
133
24438
XM_017010612.1
XP_016866101.1
132
24362
XM_017010613.1
XP_016866102.1
134
24731
XM_017010614.1
XP_016866103.1
133
24578
XM_017010615.1
XP_016866104.1
133
24578
XM_017010616.1
XP_016866105.1
134
24734
XM_017010617.1
XP_016866106.1
134
24731
XM_017010618.1
XP_016866107.1
134
24719
XM_017010619.1
XP_016866108.1
120
22813
GenBank mRNA
AB018339.2
-
22
3454
AF444779.1
-
9
1327
AF495910.1
-
134
24817
AF535142.1
-
133
24439
AK122691.1
-
14
2097
AK127076.1
-
18
2794
AK304825.1
-
8
1019
AK316101.1
-
7
1034
AY061755.1
-
49
7907
AY061756.1
-
6
897
AY184203.1
-
5
762
AY184206.1
-
47
7802
BC090927.1
-
7
1079
BX537837.1
-
16
2395
BX647837.1
-
19
2989
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0