Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HBA1 SIRNA Assays ›
          Gene Symbol:
          Hba1 Hba2
          Gene Name:
          hemoglobin, alpha 1
          hemoglobin, alpha 2
          Gene Aliases:
          HBAM, Hba-a1, Hba-a2
          -
          Chromosome Location:
          Chr.10: 15602794 - 15603649 on Build Rnor_6.0
          Chromosome Location:
          Chr.10: 15589364 - 15590207 on Build Rnor_6.0
          UniGene ID:
          Rn.107334
          Rn.203003
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s236164

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hba1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013096.1 NP_037228.1 3 421
          GenBank mRNA BC059150.1 - 3 420 6918464
          BC091567.1 - 408 7458326
          BC160818.1 - 3 404 7375173
          FQ209733.1 - 3 421
          FQ209744.1 - 3 421
          FQ209909.1 - 3 421
          FQ209912.1 - 3 421
          FQ210001.1 - 3 421
          FQ210150.1 - 3 419
          FQ210186.1 - 3 419
          FQ210277.1 - 3 419
          FQ210349.1 - 3 419
          FQ210635.1 - 3 421
          FQ210643.1 - 3 419
          FQ210690.1 - 3 420
          FQ210723.1 - 3 419
          FQ210862.1 - 3 419
          FQ210866.1 - 3 419
          FQ211077.1 - 3 421
          FQ211217.1 - 3 419
          FQ211220.1 - 3 421
          FQ211232.1 - 3 420
          FQ211239.1 - 3 419
          FQ211351.1 - 3 419
          FQ211374.1 - 3 419
          FQ211431.1 - 3 419
          FQ211513.1 - 3 419
          FQ211520.1 - 3 419
          FQ211755.1 - 3 419
          FQ211911.1 - 3 419
          FQ211972.1 - 3 419
          FQ212025.1 - 3 421
          FQ212144.1 - 3 421
          FQ212153.1 - 3 421
          FQ212186.1 - 3 419
          FQ212232.1 - 3 421
          FQ212457.1 - 3 419
          FQ212854.1 - 3 419
          FQ213301.1 - 3 419
          FQ214066.1 - 3 419
          FQ214223.1 - 3 419
          FQ216243.1 - 3 419
          FQ217723.1 - 3 419
          FQ217724.1 - 3 418
          FQ217988.1 - 3 421
          FQ218146.1 - 419
          FQ218257.1 - 3 475
          FQ218612.1 - 3 418
          FQ218968.1 - 3 421
          FQ219715.1 - 3 419
          FQ220786.1 - 3 419
          FQ221656.1 - 3 420
          FQ221912.1 - 3 421
          FQ221952.1 - 3 419
          FQ222431.1 - 3 419
          FQ222454.1 - 3 419
          FQ222672.1 - 3 419
          FQ222750.1 - 3 421
          FQ223017.1 - 3 420
          FQ224581.1 - 3 419
          FQ225427.1 - 3 420
          FQ225429.1 - 3 421
          FQ225440.1 - 3 420
          FQ225452.1 - 3 419
          FQ225777.1 - 3 419
          FQ225808.1 - 3 422
          FQ226118.1 - 3 421
          FQ226173.1 - 3 420
          FQ226203.1 - 3 419
          FQ226615.1 - 3 419
          FQ227082.1 - 3 419
          FQ228301.1 - 3 419
          FQ228436.1 - 419
          FQ230483.1 - 3 421
          FQ230901.1 - 3 419
          FQ231201.1 - 3 411
          FQ231290.1 - 3 419
          FQ232272.1 - 3 419
          FQ232481.1 - 3 419
          FQ234683.1 - 3 419
          M17083.1 - 3 421
          Gene Symbol: Hba2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007722.1 NP_001007723.1 421
          GenBank mRNA FQ209950.1 - 3 419
          FQ210108.1 - 3 419
          FQ210256.1 - 3 419
          FQ210264.1 - 3 419
          FQ210372.1 - 3 424
          FQ210420.1 - 3 418
          FQ210570.1 - 3 419
          FQ210694.1 - 3 419
          FQ211180.1 - 3 419
          FQ211270.1 - 3 419
          FQ211366.1 - 3 419
          FQ211672.1 - 3 419
          FQ211887.1 - 3 419
          FQ213126.1 - 3 419
          FQ214258.1 - 3 419
          FQ215087.1 - 3 419
          FQ217874.1 - 3 419
          FQ218140.1 - 3 421
          FQ218297.1 - 3 425
          FQ218784.1 - 3 418
          FQ221177.1 - 3 419
          FQ221489.1 - 3 419
          FQ222342.1 - 3 419
          FQ222639.1 - 3 418
          FQ222693.1 - 3 417
          FQ223754.1 - 3 421
          FQ223838.1 - 3 445
          FQ224218.1 - 3 428
          FQ224339.1 - 3 421
          FQ224679.1 - 3 419
          FQ225311.1 - 3 419
          FQ225451.1 - 3 419
          FQ225711.1 - 3 419
          FQ225789.1 - 3 421
          FQ226185.1 - 3 419
          FQ226226.1 - 574
          FQ226238.1 - 3 409
          FQ226556.1 - 3 419
          FQ226574.1 - 3 419
          FQ226576.1 - 3 419
          FQ226594.1 - 3 419
          FQ226619.1 - 3 419
          FQ226919.1 - 3 417
          FQ226927.1 - 3 419
          FQ228017.1 - 3 421
          FQ228292.1 - 3 419
          FQ228298.1 - 3 419
          FQ228306.1 - 3 419
          FQ228588.1 - 3 419
          FQ229556.1 - 3 419
          FQ230499.1 - 3 419
          FQ230501.1 - 3 419
          FQ230536.1 - 3 419
          FQ230819.1 - 3 421
          FQ230868.1 - 3 419
          FQ230877.1 - 3 421
          FQ231222.1 - 3 419
          FQ231239.1 - 3 414
          FQ231246.1 - 3 419
          FQ231250.1 - 3 419
          FQ231268.1 - 3 418
          FQ231572.1 - 3 419
          FQ231902.1 - 3 419
          FQ232213.1 - 3 419
          FQ234805.1 - 3 419
          FQ235117.1 - 3 420

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_008218, AK171893, AK010923, AK010933, AK011116, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          oxygen transport
          negative regulation of blood pressure
          protein heterooligomerization
          regulation of sensory perception of pain
          in utero embryonic development
          positive regulation of cell death
          response to hydrogen peroxide
          hydrogen peroxide catabolic process
          erythrocyte development
          oxidation-reduction process
          beta-amyloid binding
          oxygen transporter activity
          iron ion binding
          protein binding
          oxygen binding
          heme binding
          organic acid binding
          peroxidase activity
          haptoglobin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0