Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAMK2B SIRNA Assays ›
          Gene Symbol:
          CAMK2B
          Gene Name:
          calcium/calmodulin dependent protein kinase II beta
          Gene Aliases:
          CAM2, CAMK2, CAMKB
          Chromosome Location:
          Chr.7: 44217150 - 44325994 on Build GRCh38
          UniGene ID:
          Hs.351887
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s2363

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CAMK2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001220.4 NP_001211.3 6 587
          NM_001293170.1 NP_001280099.1 6 587
          NM_172078.2 NP_742075.1 6 587
          NM_172079.2 NP_742076.1 6 587
          NM_172080.2 NP_742077.1 6 587
          NM_172081.2 NP_742078.1 6 587
          NM_172082.2 NP_742079.1 6 587
          NM_172083.2 NP_742080.1 6 587
          NM_172084.2 NP_742081.1 6 587
          XM_005249862.3 XP_005249919.1 6 463
          XM_005249864.3 XP_005249921.1 6 463
          XM_006715781.2 XP_006715844.1 6 463
          XM_006715784.2 XP_006715847.1 6 463
          XM_011515547.1 XP_011513849.1 6 463
          XM_011515549.1 XP_011513851.1 6 463
          XM_011515550.1 XP_011513852.1 6 463
          XM_011515551.1 XP_011513853.1 6 463
          XM_011515552.1 XP_011513854.1 6 463
          XM_011515553.1 XP_011513855.1 6 463
          XM_011515554.1 XP_011513856.1 6 463
          XM_011515555.1 XP_011513857.1 6 463
          XM_011515556.1 XP_011513858.1 6 463
          XM_011515557.1 XP_011513859.1 6 463
          XM_011515558.2 XP_011513860.1 6 463
          XM_011515559.1 XP_011513861.1 6 372
          XM_017012660.1 XP_016868149.1 6 463
          XM_017012661.1 XP_016868150.1 6 463
          XM_017012662.1 XP_016868151.1 6 463
          XM_017012663.1 XP_016868152.1 6 372
          XM_017012664.1 XP_016868153.1 6 463
          XM_017012665.1 XP_016868154.1 6 463
          XM_017012666.1 XP_016868155.1 6 372
          XM_017012667.1 XP_016868156.1 6 372
          XM_017012668.1 XP_016868157.1 6 372
          XM_017012669.1 XP_016868158.1 6 372
          XM_017012670.1 XP_016868159.1 6 372
          GenBank mRNA AA399393.1 - 2 84
          AF078803.1 - 6 380
          AF081572.1 - 6 380
          AF081924.1 - 6 380
          AF083419.1 - 6 380
          AF112471.1 - 6 423
          AF112472.1 - 6 423
          AF140350.1 - 6 380
          AJ252236.1 - 6 464
          AJ252237.1 - 6 387
          AJ252238.1 - 6 377
          AK222703.1 - 6 411
          AK290148.1 - 6 452
          AK294138.1 - 4 310
          AK315663.1 - 6 494
          BC019070.2 - 6 581 5014712
          BX445032.2 - 6 396
          DA069361.1 - 5 239
          DB175099.1 - 6 365
          U23460.1 - 6 452

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          protein phosphorylation
          signal transduction
          positive regulation of neuron projection development
          regulation of skeletal muscle adaptation
          positive regulation of phospholipase A2 activity
          positive regulation of GTPase activity
          protein autophosphorylation
          regulation of long-term neuronal synaptic plasticity
          cell projection morphogenesis
          regulation of synapse structural plasticity
          regulation of calcium ion transport
          interferon-gamma-mediated signaling pathway
          regulation of dendritic spine development
          positive regulation of dendritic spine morphogenesis
          positive regulation of synapse maturation
          regulation of cellular response to heat
          actin binding
          protein serine/threonine kinase activity
          calmodulin-dependent protein kinase activity
          Ras guanyl-nucleotide exchange factor activity
          protein binding
          calmodulin binding
          ATP binding
          protein kinase binding
          protein homodimerization activity
          phospholipase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nbkcc:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0