Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107375.2 | NP_001100845.1 | 8 | 1787 |
Gene Symbol: LOC100911356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751804.3 | XP_003751852.1 | 8 | 1780 | ||
XM_008772208.1 | XP_008770430.1 | 8 | 1800 |