Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cyp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134844.1 | NP_001128316.1 | 1701 | |||
XM_008765492.1 | XP_008763714.1 | 1720 | ||||
XM_008765493.1 | XP_008763715.1 | 2106 | ||||
GenBank mRNA | BC166502.1 | - | 1701 | 7122207 |