Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRNDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308963.1 | NP_001295892.1 | 5 | 580 | ||
NR_034105.3 | 5 | 798 | ||||
NR_034106.2 | 5 | 663 | ||||
NR_110453.1 | 6 | 885 | ||||
NR_110454.1 | 5 | 591 | ||||
XM_017023562.1 | XP_016879051.1 | 5 | 412 | |||
XM_017023563.1 | XP_016879052.1 | 5 | 1713 | |||
GenBank mRNA | AF275804.1 | - | 5 | 406 | ||
CV575277.1 | - | 5 | 529 | |||
FJ466685.1 | - | 5 | 436 | |||
FJ466686.1 | - | 5 | 353 |