Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PINK1 SIRNA Assays ›
Gene Symbol:
PINK1
Gene Name:
PTEN induced putative kinase 1
Gene Aliases:
BRPK, PARK6
Chromosome Location:
Chr.1: 20633455 - 20651511 on Build GRCh38
UniGene ID:
Hs.389171
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s239075
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PINK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_032409.2
NP_115785.1
2
563
GenBank mRNA
AB053323.1
-
2
563
AF316873.1
-
2
539
BC028215.1
-
2
478
5214483
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
mitophagy
positive regulation of protein phosphorylation
regulation of oxidative phosphorylation
protein phosphorylation
ubiquitin-dependent protein catabolic process
response to stress
response to oxidative stress
mitochondrion organization
regulation of hydrogen peroxide metabolic process
negative regulation of gene expression
regulation of mitochondrion organization
positive regulation of peptidase activity
macroautophagy
positive regulation of macroautophagy
negative regulation of macroautophagy
phosphorylation
protein ubiquitination
peptidyl-serine phosphorylation
respiratory electron transport chain
regulation of protein ubiquitination
positive regulation of protein ubiquitination
activation of protein kinase B activity
positive regulation of synaptic transmission, dopaminergic
positive regulation of peptidyl-serine phosphorylation
positive regulation of dopamine secretion
cellular response to oxidative stress
positive regulation of protein dephosphorylation
intracellular signal transduction
peptidyl-serine autophosphorylation
TORC2 signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
regulation of protein complex assembly
negative regulation of neuron apoptotic process
positive regulation of translation
negative regulation of JNK cascade
protein stabilization
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of ubiquitin-protein transferase activity
regulation of mitochondrial membrane potential
positive regulation of protein kinase B signaling
regulation of proteasomal protein catabolic process
cellular response to hypoxia
establishment of protein localization to mitochondrion
maintenance of protein location in mitochondrion
positive regulation of mitochondrial fission
positive regulation of release of cytochrome c from mitochondria
negative regulation of mitochondrial fission
cellular response to toxic substance
mitophagy in response to mitochondrial depolarization
regulation of synaptic vesicle transport
negative regulation of autophagosome assembly
positive regulation of mitochondrial electron transport, NADH to ubiquinone
regulation of mitophagy
negative regulation of mitophagy
negative regulation of oxidative stress-induced cell death
negative regulation of oxidative stress-induced neuron death
regulation of protein targeting to mitochondrion
negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway
negative regulation of hydrogen peroxide-induced neuron intrinsic apoptotic signaling pathway
negative regulation of intrinsic apoptotic signaling pathway in response to hydrogen peroxide
positive regulation of cristae formation
positive regulation of protein targeting to mitochondrion
positive regulation of free ubiquitin chain polymerization
regulation of reactive oxygen species metabolic process
negative regulation of reactive oxygen species metabolic process
positive regulation of ATP biosynthetic process
magnesium ion binding
protease binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
calcium-dependent protein kinase activity
kinase activity
peptidase activator activity
ubiquitin protein ligase binding
protein kinase B binding
C3HC4-type RING finger domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0