Search
Search

| Gene Symbol: CAPZA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006135.2 | NP_006126.1 | 8 | 1018 | ||
| XM_017002424.1 | XP_016857913.1 | 8 | 762 | |||
| GenBank mRNA | AK223226.1 | - | 8 | 639 | ||
| AK289654.1 | - | 8 | 661 | |||
| BC000144.1 | - | 8 | 643 | 3047937 | ||
| BT019364.1 | - | 8 | 626 | |||
| BX648738.1 | - | 8 | 1309 | |||
| CR407657.1 | - | 8 | 626 | |||
| CR541819.1 | - | 8 | 626 | |||
| U56637.1 | - | 8 | 626 | |||
| Gene Symbol: BAZ2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX647250.1 | - | 4 | 2533 | ||