Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPIDR SIRNA Assays ›
Gene Symbol:
SPIDR
Gene Name:
scaffolding protein involved in DNA repair
Gene Aliases:
KIAA0146
Chromosome Location:
Chr.8: 47260878 - 47736001 on Build GRCh38
UniGene ID:
Hs.381058
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23975
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SPIDR
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080394.3
NP_001073863.1
3
288
NM_001282919.1
NP_001269848.1
3
251
NR_104581.1
3
288
XM_005251189.3
XP_005251246.1
3
230
XM_005251191.3
XP_005251248.1
3
230
XM_005251193.3
XP_005251250.1
3
230
XM_005251195.4
XP_005251252.1
3
238
XM_005251198.4
XP_005251255.1
3
259
XM_005251199.4
XP_005251256.1
2
174
XM_006716443.3
XP_006716506.1
3
198
XM_006716444.3
XP_006716507.1
3
222
XM_011517497.2
XP_011515799.1
3
230
XM_011517500.2
XP_011515802.1
3
248
XM_017013268.1
XP_016868757.1
3
230
XM_017013269.1
XP_016868758.1
3
230
XM_017013270.1
XP_016868759.1
3
248
XM_017013271.1
XP_016868760.1
3
345
XM_017013273.1
XP_016868762.1
3
259
XM_017013274.1
XP_016868763.1
3
238
XM_017013277.1
XP_016868766.1
3
215
XM_017013278.1
XP_016868767.1
3
204
XM_017013279.1
XP_016868768.1
2
174
XM_017013280.1
XP_016868769.1
3
259
XM_017013281.1
XP_016868770.1
3
224
XM_017013282.1
XP_016868771.1
3
238
XM_017013283.1
XP_016868772.1
2
174
XM_017013284.1
XP_016868773.1
3
238
XM_017013285.1
XP_016868774.1
3
259
XM_017013286.1
XP_016868775.1
2
174
XM_017013287.1
XP_016868776.1
3
224
XM_017013290.1
XP_016868779.1
2
174
XM_017013291.1
XP_016868780.1
2
174
GenBank mRNA
AK122820.1
-
4
586
AK293809.1
-
3
211
AK293972.1
-
3
211
AK297681.1
-
3
210
AK303579.1
-
3
198
AK309842.1
-
3
224
BC031627.1
-
3
213
D63480.1
-
3
216
DR000775.1
-
3
277
HY012281.1
-
3
289
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
cellular response to DNA damage stimulus
regulation of double-strand break repair via homologous recombination
positive regulation of protein complex assembly
regulation of establishment of protein localization to chromosome
cellular response to ionizing radiation
cellular response to hydroxyurea
cellular response to camptothecin
positive regulation of double-strand break repair
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-gs8n8:80/10.239.163.223:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0