Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM1 SIRNA Assays ›
Gene Symbol:
R3HDM1
Gene Name:
R3H domain containing 1
Gene Aliases:
R3HDM
Chromosome Location:
Chr.2: 135531429 - 135725270 on Build GRCh38
UniGene ID:
Hs.412462
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23987
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282798.1
NP_001269727.1
18, 19
2491
NM_001282799.1
NP_001269728.1
15, 16
2099
NM_001282800.1
NP_001269729.1
15, 16
2148
NM_015361.3
NP_056176.2
18, 19
2483
XM_005263626.3
XP_005263683.1
18, 19
2289
XM_005263627.3
XP_005263684.1
18, 19
2286
XM_005263628.3
XP_005263685.1
17, 18
2220
XM_005263629.3
XP_005263686.1
17, 18
2217
XM_006712389.2
XP_006712452.1
18, 19
2475
XM_011510870.1
XP_011509172.1
19, 20
2577
XM_011510871.1
XP_011509173.1
19, 20
2593
XM_011510872.2
XP_011509174.1
18, 19
2535
XM_011510873.2
XP_011509175.1
19, 20
2529
XM_011510874.2
XP_011509176.1
17, 18
2445
XM_011510875.2
XP_011509177.1
17, 18
2461
XM_011510876.2
XP_011509178.1
19, 20
2391
XM_011510877.2
XP_011509179.1
19, 20
2343
XM_011510878.2
XP_011509180.1
18, 19
2322
XM_011510879.2
XP_011509181.1
18, 19
2274
XM_011510881.1
XP_011509183.1
19, 20
2577
XM_017003718.1
XP_016859207.1
18, 19
2488
XM_017003719.1
XP_016859208.1
17, 18
2467
XM_017003720.1
XP_016859209.1
18, 19
2440
XM_017003721.1
XP_016859210.1
19, 20
2407
XM_017003722.1
XP_016859211.1
16, 17
2343
XM_017003723.1
XP_016859212.1
16, 17
2359
XM_017003724.1
XP_016859213.1
16, 17
2178
XM_017003725.1
XP_016859214.1
20, 21
2669
XM_017003726.1
XP_016859215.1
20, 21
2739
XM_017003727.1
XP_016859216.1
19, 20
2697
XM_017003728.1
XP_016859217.1
17, 18
2172
XM_017003729.1
XP_016859218.1
18, 19
2514
XM_017003730.1
XP_016859219.1
16, 17
2258
XM_017003731.1
XP_016859220.1
19, 20
2588
XM_017003732.1
XP_016859221.1
19, 20
2563
XM_017003733.1
XP_016859222.1
20, 21
2712
XM_017003734.1
XP_016859223.1
21, 22
2872
XM_017003735.1
XP_016859224.1
18, 19
2470
XM_017003736.1
XP_016859225.1
17, 18
2300
GenBank mRNA
AK127793.1
-
15, 16
2119
AK290015.1
-
18, 19
2377
AK297000.1
-
10, 11
1269
BC041093.1
-
18, 19
2425
5556247
D21852.1
-
15, 16
2053
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0