Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAST SIRNA Assays ›
Gene Symbol:
CAST
Gene Name:
calpastatin
Gene Aliases:
BS-17, PLACK
Chromosome Location:
Chr.5: 96662037 - 96774683 on Build GRCh38
UniGene ID:
Hs.436186
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2399
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAST
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042440.3
NP_001035905.1
24
2101
NM_001190442.1
NP_001177371.1
25
1973
NM_001284212.2
NP_001271141.1
22
1749
NM_001284213.2
NP_001271142.1
21
1659
NM_173060.3
NP_775083.1
23
1809
NR_104285.1
11
962
XM_006714696.3
XP_006714759.1
27
2115
XM_006714697.3
XP_006714760.1
26
2129
XM_006714698.3
XP_006714761.1
25
2093
XM_006714699.3
XP_006714762.1
26
2056
XM_006714700.3
XP_006714763.1
25
2075
XM_006714701.3
XP_006714764.1
25
2091
XM_006714702.3
XP_006714765.1
25
1967
XM_006714703.3
XP_006714766.1
24
2052
XM_006714704.3
XP_006714767.1
25
2000
XM_006714705.3
XP_006714768.1
24
2006
XM_006714706.3
XP_006714769.1
24
1927
XM_006714707.3
XP_006714770.1
24
1878
XM_006714708.3
XP_006714771.1
23
1838
XM_006714709.3
XP_006714772.1
23
1801
XM_006714710.3
XP_006714773.1
22
1756
XM_006714711.3
XP_006714774.1
22
1801
XM_006714712.3
XP_006714775.1
21
1748
XM_006714713.3
XP_006714776.1
11
965
XM_006714714.3
XP_006714777.1
11
962
XM_006714715.3
XP_006714778.1
12
1691
XM_011543654.2
XP_011541956.1
25
2051
XM_011543655.2
XP_011541957.1
25
2048
XM_011543656.2
XP_011541958.1
24
1995
XM_011543657.2
XP_011541959.1
24
1877
XM_011543658.2
XP_011541960.1
22
1730
XM_017009911.1
XP_016865400.1
25
2090
XM_017009912.1
XP_016865401.1
26
2101
XM_017009913.1
XP_016865402.1
26
2073
XM_017009914.1
XP_016865403.1
25
2072
XM_017009915.1
XP_016865404.1
26
2077
XM_017009916.1
XP_016865405.1
25
2088
XM_017009917.1
XP_016865406.1
25
2009
XM_017009918.1
XP_016865407.1
24
2036
XM_017009919.1
XP_016865408.1
24
2009
XM_017009920.1
XP_016865409.1
24
1919
XM_017009921.1
XP_016865410.1
24
1933
XM_017009922.1
XP_016865411.1
23
1970
XM_017009923.1
XP_016865412.1
23
1967
XM_017009924.1
XP_016865413.1
23
1949
XM_017009925.1
XP_016865414.1
23
1888
XM_017009926.1
XP_016865415.1
23
1885
XM_017009927.1
XP_016865416.1
24
1874
XM_017009928.1
XP_016865417.1
23
1873
XM_017009929.1
XP_016865418.1
23
1792
XM_017009930.1
XP_016865419.1
22
1762
XM_017009931.1
XP_016865420.1
22
1753
XM_017009932.1
XP_016865421.1
21
1816
GenBank mRNA
AB208816.1
-
25
1937
AF327443.1
-
20
1651
AK292289.1
-
11
965
AK292356.1
-
11
938
AK296919.1
-
25
1931
AK297879.1
-
23
1751
AK298541.1
-
24
1878
AK299384.1
-
22
1509
AK299402.1
-
24
1858
AK300624.1
-
21
1642
AK302206.1
-
12
965
AK311598.1
-
12
1019
AK314290.1
-
22
1754
AK315146.1
-
22
1753
AK315673.1
-
11
910
AL045308.1
-
4
241
AL832174.1
-
23
1794
AL832349.1
-
23
1797
AY189741.1
-
13
964
BC013579.1
-
24
1804
3878564
BC078149.1
-
19
1363
BT009783.1
-
21
1586
BX648397.1
-
22
1700
D16217.1
-
24
1871
D50827.1
-
2
119
S73329.1
-
3
143
U26724.2
-
15
1234
U31345.1
-
13
978
U31346.1
-
3
194
U38525.1
-
6
437
U58996.2
-
11
941
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
liver development
egg activation
brain development
myoblast fusion
aging
negative regulation of endopeptidase activity
organ regeneration
myoblast differentiation
negative regulation of cell cycle arrest
cell-cell adhesion
negative regulation of type B pancreatic cell apoptotic process
protease binding
endopeptidase inhibitor activity
protein binding
calcium-dependent cysteine-type endopeptidase inhibitor activity
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-mtn5c:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0