Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L13 SIRNA Assays ›
Gene Symbol:
BCL2L13
Gene Name:
BCL2 like 13
Gene Aliases:
BCL-RAMBO, Bcl2-L-13, MIL1
Chromosome Location:
Chr.22: 17628855 - 17730855 on Build GRCh38
UniGene ID:
Hs.631672
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24397
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L13
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001270726.1
NP_001257655.1
2
322
NM_001270727.1
NP_001257656.1
2
322
NM_001270729.1
NP_001257658.1
3
493
NM_001270730.1
NP_001257659.1
2
322
NM_001270731.1
NP_001257660.1
3
493
NM_001270732.1
NP_001257661.1
3
493
NM_001270734.1
NP_001257663.1
3
493
NM_001270735.1
NP_001257664.1
3
493
NM_015367.3
NP_056182.2
3
493
NR_073068.1
3
305
NR_073069.1
2
190
XM_011546119.1
XP_011544421.1
3
856
XM_011546120.1
XP_011544422.1
3
858
XM_011546121.2
XP_011544423.1
4
841
XM_011546123.1
XP_011544425.1
4
435
XM_017028725.1
XP_016884214.1
3
856
XM_017028726.1
XP_016884215.1
5
1142
XM_017028727.1
XP_016884216.1
4
435
XM_017028728.1
XP_016884217.1
4
1027
XM_017028729.1
XP_016884218.1
4
435
XM_017028730.1
XP_016884219.1
2
149
XM_017028731.1
XP_016884220.1
3
856
XM_017028732.1
XP_016884221.1
2
169
XM_017028733.1
XP_016884222.1
3
493
XM_017028734.1
XP_016884223.1
4
1027
GenBank mRNA
AF246665.1
-
4
879
AF325209.1
-
2
140
AF539453.1
-
2
140
AK291449.1
-
3
313
AK314489.1
-
3
320
BC007658.2
-
3
252
3343994
BE900458.1
-
3
274
BX354578.2
-
3
358
BX407004.2
-
2
75
CN420914.1
-
3
287
CR456521.1
-
3
396
DA154984.1
-
2
149
DA478149.1
-
2
152
DA799358.1
-
3
495
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
protein binding
cysteine-type endopeptidase activator activity involved in apoptotic process
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0