Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2461
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
8
1220
NM_001198626.1
NP_001185555.1
6
1078
NM_001198627.1
NP_001185556.1
7
1138
NM_001198628.1
NP_001185557.1
5
735
NM_001198629.1
NP_001185558.1
5
861
NM_001198630.1
NP_001185559.1
6
921
NM_001198631.1
NP_001185560.1
5
934
NM_001198632.1
NP_001185561.1
5
852
NM_001198633.1
NP_001185562.1
4
823
NM_001198634.1
NP_001185563.1
4
761
NM_001198679.1
NP_001185608.1
5
905
NM_004349.3
NP_004340.1
4
871
NM_175634.2
NP_783552.1
5
996
NM_175635.2
NP_783553.1
4
688
NM_175636.2
NP_783554.1
3
770
XM_006716676.3
XP_006716739.1
6
805
XM_011517351.2
XP_011515653.1
6
850
XM_011517352.2
XP_011515654.1
6
1090
XM_011517353.2
XP_011515655.1
5
798
XM_017013930.1
XP_016869419.1
5
1106
XM_017013931.1
XP_016869420.1
4
628
XM_017013932.1
XP_016869421.1
6
993
XM_017013933.1
XP_016869422.1
7
1057
XM_017013934.1
XP_016869423.1
7
1165
XM_017013935.1
XP_016869424.1
6
970
XM_017013936.1
XP_016869425.1
5
1064
XM_017013937.1
XP_016869426.1
6
890
XM_017013938.1
XP_016869427.1
5
754
GenBank mRNA
AK297616.1
-
4
585
AK312592.1
-
8
982
BC005850.1
-
4
650
2961112
BC067078.1
-
4
641
2964656
BT009871.1
-
3
430
CR456792.1
-
3
460
D14289.1
-
3
727
D14821.1
-
4
625
D43638.1
-
4
871
KF946505.1
-
4
489
KF946507.1
-
4
489
KF946508.1
-
4
489
KF946510.1
-
4
489
KF946511.1
-
4
475
KF946512.1
-
5
1779
KF946513.1
-
4
2159
KF946514.1
-
4
475
KF946515.1
-
3
715
KF946516.1
-
3
715
KF946517.1
-
3
715
KF946518.1
-
3
715
X79990.1
-
3
453
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0