Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LETMD1 SIRNA Assays ›
Gene Symbol:
LETMD1
Gene Name:
LETM1 domain containing 1
Gene Aliases:
1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
Chromosome Location:
Chr.12: 51048230 - 51068984 on Build GRCh38
UniGene ID:
Hs.655272
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s24659
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LETMD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243689.1
NP_001230618.1
8
1036
NM_001300765.1
NP_001287694.1
5
625
NM_015416.4
NP_056231.3
8
997
NR_045017.1
8
872
NR_045018.1
7
881
NR_045019.1
6
798
NR_045020.1
4
509
XM_006719335.1
XP_006719398.1
7
819
XM_006719336.1
XP_006719399.1
7
707
XM_006719339.1
XP_006719402.1
8
935
XM_006719340.1
XP_006719403.1
7
756
XM_011538161.2
XP_011536463.1
8
1004
XM_011538164.2
XP_011536466.1
7
854
XM_011538165.2
XP_011536467.1
5
632
XM_011538167.1
XP_011536469.1
3
426
XM_017019151.1
XP_016874640.1
8
1006
XM_017019152.1
XP_016874641.1
8
972
XM_017019153.1
XP_016874642.1
7
856
XM_017019154.1
XP_016874643.1
7
819
XM_017019155.1
XP_016874644.1
8
844
XM_017019156.1
XP_016874645.1
6
704
XM_017019157.1
XP_016874646.1
6
666
XM_017019158.1
XP_016874647.1
7
707
XM_017019159.1
XP_016874648.1
5
635
XM_017019160.1
XP_016874649.1
5
597
XM_017019161.1
XP_016874650.1
5
603
XM_017019162.1
XP_016874651.1
5
573
XM_017019163.1
XP_016874652.1
6
666
XM_017019164.1
XP_016874653.1
7
873
XM_017019165.1
XP_016874654.1
8
935
XM_017019166.1
XP_016874655.1
7
756
GenBank mRNA
AF195651.2
-
8
969
AF315598.1
-
8
833
AF329277.1
-
7
880
AK000639.1
-
8
984
AK057866.1
-
8
942
AK123080.1
-
1
583
AK127540.1
-
8
997
AK225836.1
-
8
955
AK301955.1
-
8
872
AK315119.1
-
8
964
AL050286.1
-
3
489
AY259835.1
-
6
740
AY259836.1
-
4
451
BC000395.2
-
3
246
BC019274.1
-
3
246
BC064943.1
-
8
943
6009854
BM549678.1
-
481
BX343550.2
-
6
739
BX376670.2
-
5
566
Back To Top
More Information
Panther Classification:
Molecular Function -
secondary carrier transporter
Gene Ontology Categories:
Process(es)
protein binding
ribosome binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0