Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PAMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001991.2 | NP_001001991.1 | 9 | 1562 | ||
NM_001282675.1 | NP_001269604.1 | 11 | 1552 | |||
NM_001282676.1 | NP_001269605.1 | 7 | 1229 | |||
NM_015430.3 | NP_056245.2 | 10 | 1613 | |||
GenBank mRNA | AF370388.1 | - | 8 | 1051 | ||
AK027841.1 | - | 10 | 1213 | |||
AK297092.1 | - | 11 | 1553 | |||
AK300185.1 | - | 7 | 814 | |||
AK316188.1 | - | 7 | 801 | |||
AL832391.1 | - | 9 | 1128 | |||
AY358346.1 | - | 9 | 1141 | |||
BC089434.1 | - | 10 | 1205 | 30558617 | ||
BX640676.1 | - | 9 | 1562 |