Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RCHY1 SIRNA Assays ›
          Gene Symbol:
          RCHY1
          Gene Name:
          ring finger and CHY zinc finger domain containing 1
          Gene Aliases:
          ARNIP, CHIMP, PIRH2, PRO1996, RNF199, ZCHY, ZNF363
          Chromosome Location:
          Chr.4: 75479037 - 75515057 on Build GRCh38
          UniGene ID:
          Hs.48297
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s24704

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RCHY1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001009922.2 NP_001009922.1 2 306
          NM_001278538.1 NP_001265467.1 2 262
          NM_001278539.1 NP_001265468.1 2 348
          NM_015436.3 NP_056251.2 2 306
          NR_037913.1 2 306
          NR_037914.1 2 306
          NR_103723.1 2 306
          NR_103724.1 2 306
          NR_103725.1 1 84
          XM_011531838.2 XP_011530140.1 2 279
          XM_011531839.2 XP_011530141.1 2 467
          GenBank mRNA AB209072.1 - 2 169
          AF247041.1 - 2 162
          AF255666.1 - 2 188
          AF305424.1 - 2 196
          AK091501.1 - 2 277
          AK309555.1 - 2 153
          AL050144.1 - 1 65
          AY888047.1 - 2 188
          BC047393.1 - 2 306 5301408
          BF102905.1 - 1 84
          BG703058.1 - 2 179
          BQ959587.1 - 2 135
          DA197338.1 - 2 262
          GQ250944.1 - 2 162
          GQ250945.1 - 2 162
          GQ250946.1 - 2 162
          GU937000.1 - 2 189
          JF691538.1 - 2 162
          JF691539.1 - 2 162

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein ubiquitination
          positive regulation of protein ubiquitination
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          error-free translesion synthesis
          p53 binding
          ubiquitin-protein transferase activity
          receptor binding
          protein binding
          zinc ion binding
          ligase activity
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0