Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFBR3 SIRNA Assays ›
          Gene Symbol:
          TGFBR3
          Gene Name:
          transforming growth factor beta receptor 3
          Gene Aliases:
          BGCAN, betaglycan
          Chromosome Location:
          Chr.1: 91680343 - 91906002 on Build GRCh38
          UniGene ID:
          Hs.482390
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s25

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TGFBR3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195683.1 NP_001182612.1 11 2149
          NM_001195684.1 NP_001182613.1 12 1985
          NM_003243.4 NP_003234.2 11 2152
          NR_036634.1 12 2249
          XM_006710867.2 XP_006710930.1 11 1871
          XM_006710868.2 XP_006710931.1 11 1837
          GenBank mRNA AJ251961.1 - 10 1699
          AK225799.1 - 11 1927
          AK291345.1 - 11 1793
          BC099914.1 - 14 2388
          BC126116.1 - 11 1805 8991832
          BC136295.1 - 11 1805 9020282
          BC143869.1 - 12 1902
          HG764816.1 - 12 1985
          L07594.1 - 12 1979

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          epithelial to mesenchymal transition
          liver development
          heart morphogenesis
          immune response
          transforming growth factor beta receptor signaling pathway
          transforming growth factor beta receptor complex assembly
          cell growth
          cell migration
          BMP signaling pathway
          positive regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          organ regeneration
          response to follicle-stimulating hormone
          response to prostaglandin E
          response to luteinizing hormone
          intracellular signal transduction
          regulation of protein binding
          negative regulation of epithelial cell proliferation
          negative regulation of cellular component movement
          ventricular cardiac muscle tissue morphogenesis
          palate development
          cardiac muscle cell proliferation
          definitive hemopoiesis
          cardiac epithelial to mesenchymal transition
          definitive erythrocyte differentiation
          heart trabecula formation
          pathway-restricted SMAD protein phosphorylation
          transforming growth factor beta-activated receptor activity
          type II transforming growth factor beta receptor binding
          transforming growth factor beta receptor binding
          protein binding
          glycosaminoglycan binding
          heparin binding
          coreceptor activity
          fibroblast growth factor binding
          PDZ domain binding
          SMAD binding
          activin binding
          transforming growth factor beta binding
          transforming growth factor beta receptor activity, type III

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0