Search Thermo Fisher Scientific
Gene Symbol: CNTNAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014141.5 | NP_054860.1 | 19, 20 | 3772 | ||
GenBank mRNA | AB020675.1 | - | 19, 20 | 3281 | ||
AB073883.1 | - | 7, 8 | 1192 | |||
AF319045.1 | - | 19, 20 | 3396 | |||
AK294098.1 | - | 4, 5 | 882 | |||
AK296532.1 | - | 8, 9 | 1428 | |||
AK315113.1 | - | 19, 20 | 3361 | |||
BC093780.1 | - | 19, 20 | 3299 | 7939625 | ||
BC113373.1 | - | 19, 20 | 3299 | 8322425 | ||
KJ534810.1 | - | 3, 4 | 799 | |||
KJ534811.1 | - | 18, 19 | 3145 | |||
KJ534812.1 | - | 355 | ||||
KJ534813.1 | - | 6, 7 | 1255 | |||
KJ535069.1 | - | 1185 | ||||
KJ535088.1 | - | 8, 9 | 1733 |