Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HEYL SIRNA Assays ›
          Gene Symbol:
          HEYL
          Gene Name:
          hes related family bHLH transcription factor with YRPW motif-like
          Gene Aliases:
          HESR3, HEY3, HRT3, bHLHb33
          Chromosome Location:
          Chr.1: 39623431 - 39639676 on Build GRCh38
          UniGene ID:
          Hs.472566
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s25474

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HEYL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014571.3 NP_055386.1 5 409
          XM_005270745.2 XP_005270802.1 5 463
          GenBank mRNA AF311885.1 - 5 358
          AJ272215.1 - 5 368
          AL541019.3 - 5 409
          BC006087.1 - 5 379 2964729

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107977.

          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          outflow tract morphogenesis
          atrioventricular valve morphogenesis
          pulmonary valve morphogenesis
          epithelial to mesenchymal transition involved in endocardial cushion formation
          endocardial cushion morphogenesis
          cardiac ventricle morphogenesis
          transcription from RNA polymerase II promoter
          Notch signaling pathway
          mesenchymal cell development
          glomerulus development
          skeletal muscle cell differentiation
          positive regulation of neuron differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cardiac epithelial to mesenchymal transition
          ventricular septum morphogenesis
          negative regulation of androgen receptor signaling pathway
          cellular response to BMP stimulus
          proximal tubule development
          negative regulation of androgen receptor activity
          RNA polymerase II core promoter sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          transcription factor activity, protein binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription corepressor activity
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          transcription factor binding
          microsatellite binding
          protein homodimerization activity
          protein heterodimerization activity
          AF-1 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0