Search
Search

TINAG| Gene Symbol: TINAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014464.3 | NP_055279.3 | 1 | 259 | ||
| XM_006715062.2 | XP_006715125.1 | 2 | 242 | |||
| XM_011514497.2 | XP_011512799.1 | 1 | 241 | |||
| XM_017010745.1 | XP_016866234.1 | 1 | 223 | |||
| XM_017010746.1 | XP_016866235.1 | 1 | 242 | |||
| XM_017010747.1 | XP_016866236.1 | 1 | 243 | |||
| GenBank mRNA | AB022277.1 | - | 1 | 114 | ||
| AF195116.1 | - | 1 | 113 | |||
| AF195117.1 | - | 1 | 113 | |||
| AK312918.1 | - | 1 | 223 | |||
| BC056235.1 | - | 1 | 223 | |||
| BC070278.1 | - | 1 | 259 | 4607306 | ||