Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HCC1L SIRNA Assays ›
Gene Symbol:
R3HCC1L
Gene Name:
R3H domain and coiled-coil containing 1 like
Gene Aliases:
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location:
Chr.10: 98134608 - 98244897 on Build GRCh38
UniGene ID:
Hs.419800
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26095
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
392.00
Your Price
Online offer:
380.65
380.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HCC1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256619.1
NP_001243548.1
5
1885
NM_001256620.1
NP_001243549.1
4
1834
NM_014472.4
NP_055287.4
4
1830
NM_138469.2
NP_612478.2
5
1927
XM_011539639.1
XP_011537941.1
5
2142
XM_011539640.1
XP_011537942.1
6
1947
XM_011539641.1
XP_011537943.1
5
1899
XM_011539642.1
XP_011537944.1
6
2239
XM_011539643.1
XP_011537945.1
4
1826
XM_011539644.1
XP_011537946.1
4
1830
XM_011539645.2
XP_011537947.1
4
2073
XM_011539646.1
XP_011537948.1
3
1705
XM_011539647.1
XP_011537949.1
3
1707
XM_011539648.1
XP_011537950.1
2
1580
XM_011539649.1
XP_011537951.1
5
2142
XM_017016074.1
XP_016871563.1
4
3038
XM_017016075.1
XP_016871564.1
2
1580
XM_017016076.1
XP_016871565.1
5
1874
XM_017016077.1
XP_016871566.1
7
2037
XM_017016078.1
XP_016871567.1
6
1959
XM_017016079.1
XP_016871568.1
6
1973
XM_017016080.1
XP_016871569.1
4
1879
XM_017016081.1
XP_016871570.1
5
1961
XM_017016082.1
XP_016871571.1
3
1753
XM_017016083.1
XP_016871572.1
4
1769
GenBank mRNA
AF525304.1
-
5
1838
AK094479.1
-
1
341
BC098268.1
-
5
1868
40026913
BC098359.1
-
4
1771
40026912
BC099848.1
-
4
1771
40026916
CN352617.1
-
1
141
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleotide binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0