Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SGSM3 SIRNA Assays ›
          Gene Symbol:
          SGSM3
          Gene Name:
          small G protein signaling modulator 3
          Gene Aliases:
          MAP, RABGAP5, RUSC3, RUTBC3, RabGAP-5, rabGAPLP
          Chromosome Location:
          Chr.22: 40370530 - 40410289 on Build GRCh38
          UniGene ID:
          Hs.474914
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s26223

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SGSM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301849.1 NP_001288778.1 12, 13 1621
          NM_015705.5 NP_056520.2 13, 14 1766
          XM_005261572.2 XP_005261629.1 13, 14 1793
          XM_005261573.3 XP_005261630.1 13, 14 1798
          XM_005261574.4 XP_005261631.1 13, 14 1798
          XM_005261575.1 XP_005261632.1 12, 13 1661
          XM_011530158.1 XP_011528460.1 12, 13 1617
          XM_011530159.1 XP_011528461.1 13, 14 1681
          XM_011530160.1 XP_011528462.1 12, 13 1598
          XM_011530161.1 XP_011528463.1 13, 14 1790
          XM_011530162.2 XP_011528464.1 13, 14 1676
          XM_017028775.1 XP_016884264.1 12, 13 1585
          XM_017028776.1 XP_016884265.1 12, 13 1654
          XM_017028777.1 XP_016884266.1 12, 13 1654
          XM_017028778.1 XP_016884267.1 11, 12 1517
          XM_017028779.1 XP_016884268.1 12, 13 1707
          XM_017028780.1 XP_016884269.1 12, 13 1696
          GenBank mRNA AB196956.1 - 11, 12 1535
          AB275763.1 - 12, 13 1639
          AB449880.1 - 11, 12 1528
          AK295469.1 - 13, 14 1712
          AK297599.1 - 12, 13 1614
          AK301042.1 - 12, 13 1534
          AY308849.1 - 13, 14 1701
          BC008078.2 - 13, 14 1692 3027777
          BC127850.1 - 6, 7 842
          CR456441.1 - 12, 13 1564

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          cell cycle arrest
          regulation of vesicle fusion
          regulation of Rab protein signal transduction
          Rap protein signal transduction
          plasma membrane to endosome transport
          activation of GTPase activity
          regulation of cilium assembly
          GTPase activator activity
          protein binding
          Rab GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0